Skip to Content
Merck
All Photos(1)

Key Documents

92325

Millipore

Bacillus ChromoSelect Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Bacillus cereus ChromoSelect Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
chromogenic mixture, 3.2 g/L
D-mannitol, 10.0 g/L
meat extract, 1.0 g/L
peptic digest of animal tissue, 10 g/L
phenol red, 0.025 g/L
sodium chloride, 10.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.1±0.2 (25)

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

selective and differential for Bacillus cereus
selective and differential for Bacillus spp.

Application

Bacillus ChromoSelect Agar is a differential medium recommended for rapid identification of Bacillus species from a mixed culture by chromogenic method.

Preparation Note

Suspend 49.2 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 45-50°C and aseptically add the rehydrated contents of 1 vial of Polymyxin B Selektiv-Supplement (Cat. No. P9602) if desired. Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service