Skip to Content
Merck
All Photos(1)

Documents

92091

Supelco

Periodic table mix 1 for ICP

TraceCERT®, 33 elements, 10 mg/L in nitric acid

Synonym(s):

ICP Elemental Table Mix 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

10 mg/L in nitric acid

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Periodic table mix 1 for ICP has been used as a standard for the quantitative determination of mineral composition in Tenualosa ilisha using Inductively coupled plasma atomic emission spectroscopy (ICP-AES).

Analysis Note

10 mg/L: Al, As, Ba, Be, Bi, B, Ca, Cd, Cs, Cr, Co, Cu, Ga, In, Fe, Pb, Li, Mg, Mn, Ni, P, K, Rb, Se, Si, Ag, Na, Sr, S, Te, Tl, V and Zn in 10% nitric acid (contains HF traces)

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hussein Kadhem Al-Hakeim et al.
Biological trace element research, 189(2), 379-386 (2018-09-02)
A considerable attention has been focused on the possible association between ultra-trace elements (UTEs) status and pathogenesis of many diseases including rheumatoid arthritis (RA). UTEs have important roles in numerous metabolic processes. Serum Cs, Rb, and Re levels in RA
Nutrigenomic studies on hilsa to evaluate flesh quality attributes and genes associated with fatty acid metabolism from the rivers Hooghly and Padma
Ganguly S, et al.
Food Research International, 103, 21-29 (2018)
Hadis Karami et al.
Biological trace element research, 199(1), 389-400 (2020-04-22)
The present study was devoted to investigate the PTE concentrations including cadmium (Cd), chromium (Cr), copper(Cu), iron (Fe), mercury (Hg), manganese (Mn), nickel (Ni), lead (Pb), arsenic(As), and zinc (Zn) among six types of edible wild, 23 cultivated mushroom samples
Nabi Shariatifar et al.
Biological trace element research, 198(2), 721-731 (2020-03-20)
Rice is one of the most valuable nutrients in the diet of most people in the world. The aim of this study was to evaluate the effect of various pre-cooking (washing, soaking) and cooking processes (traditional and rinse) of rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service