Skip to Content
Merck
All Photos(2)

Key Documents

90034

Supelco

4-Hydroxy-3-methoxycinnamic acid

mixture of isomers, analytical standard

Synonym(s):

Ferulic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O(C)c1c(ccc(c1)\C=C\C(=O)O)O

InChI

1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+

InChI key

KSEBMYQBYZTDHS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxy-3-methoxycinnamic acid is a ubiquitous plant constituent primarily occurring in seeds and leaves. It arises from the metabolism of phenylalanine and tyrosine.

Application

4-Hydroxy-3-methoxycinnamic acid may be used as a reference standard in the determination of the analyte in Chinese Danggui using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay of free ferulic acid and total ferulic acid for quality assessment of Angelica sinensis.
Lu H-G, et al.
Journal of Chromatography A, 1068(2), 209-219 (2005)
Christina E Turi et al.
Planta medica, 79(14), 1370-1379 (2013-07-24)
Ligusticum canbyi (J.M. Coult & Rose) is a medicinal understory forest species used in traditional rituals and ceremonies for spiritual enlightenment and improved mental health. Very little is known about the phytochemical complexity or diversity of L. canbyi tissues or extracts.
Yosuke Enomoto et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(9), 1055-1059 (2018-07-31)
Drosophila melanogaster is a model organism in neurodegenerative disease research. In neurodegenerative study, the direct spatial information of neurotransmitters such as γ-aminobutyric acid (GABA) in the brain of Drosophila melanogaster is important to understand the role of GABA. Matrix-assisted laser
Danika Koegelenberg et al.
Food chemistry, 221, 1606-1613 (2016-12-17)
Effects on physical properties of white bread of adding crude (E1) and partially purified (E2) arabinoxylans (AX) from wheat bran to partially replace flour during baking, were investigated to identify optimal dosage. The E1 and E2 had molecular weights of
Laura Ferrochio et al.
International journal of food microbiology, 167(2), 215-220 (2013-10-22)
The aim of this study was to evaluate the efficacy of ferulic acid (1, 10, 20 and 25 mM) at different water activity (aw) values (0.99, 0.98, 0.96 and 0.93) at 25 °C on growth and fumonisin production by Fusarium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service