Skip to Content
Merck
All Photos(1)

Key Documents

84818

Supelco

Bis(2-ethylhexyl) sebacate

Selectophore, ≥97.0%

Synonym(s):

Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
426.67
Beilstein:
1806504
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

description

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥97.0% (GC)
≥97.0%

form

liquid

refractive index

n20/D 1.450 (lit.)

bp

212 °C/1 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) sebacate (BEHS) is a plasticizer widely used in preparing ion-selective electrodes.
Visit our Sensor Applications portal to learn more.

Application

BEHS was used in preparing PVC-membrane anionic-surfactants-selective electrodes.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercury (II) ion-selective electrodes based on p-tert-butyl calix [4] crowns with imine units.
Mahajan, Rakesh Kumar, et al.
Analytical Sciences, 20.5, 811-814 (2004)
A new ion-selective electrode for anionic surfactants.
Segui, Ma Jesus, et al.
Talanta, 71.1, 333-338 (2007)
Kevin R Wilson et al.
Physical chemistry chemical physics : PCCP, 14(4), 1468-1479 (2011-12-14)
The heterogeneous reactions of hydroxyl radicals (OH) with squalane and bis(2-ethylhexyl) sebacate (BES) particles are used as model systems to examine how distributions of reaction products evolve during the oxidation of chemically reduced organic aerosol. A kinetic model of multigenerational
P J Anderson et al.
The American review of respiratory disease, 144(3 Pt 1), 649-654 (1991-09-01)
How variable is the deposition of inhaled methacholine (MCH) in the respiratory tract during a challenge test? Does this variability contribute to the variability of airway responsiveness? To examine these questions we estimated the deposition of polydisperse MCH droplets by
C S Kim et al.
Journal of applied physiology (Bethesda, Md. : 1985), 81(5), 2203-2213 (1996-11-01)
Detailed regional deposition of inhaled particles was investigated in young adults (n = 11) by use of a serial bolus aerosol delivery technique. A small bolus (45 ml half-width) of monodisperse aerosols [1-, 3-, and 5-microns particle diameter (Dp)] was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service