Skip to Content
Merck
All Photos(1)

Key Documents

83912

Sigma-Aldrich

Castor oil

tested according to Ph. Eur.

Synonym(s):

Ricini oleum virginale

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

biological source

Ricinus communis

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

form

viscous liquid

refractive index

n20/D 1.478 (lit.)

bp

313 °C (lit.)

density

0.952-0.965 g/mL at 25

lipid type

oils

SMILES string

O(C(COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)C(=O)CCCCCCC\C=C/CC(O)CCCCCC

InChI

1S/C57H104O9/c1-4-7-10-31-40-51(58)43-34-25-19-13-16-22-28-37-46-55(61)64-49-54(66-57(63)48-39-30-24-18-15-21-27-36-45-53(60)42-33-12-9-6-3)50-65-56(62)47-38-29-23-17-14-20-26-35-44-52(59)41-32-11-8-5-2/h25-27,34-36,51-54,58-60H,4-24,28-33,37-50H2,1-3H3/b

InChI key

ZEMPKEQAKRGZGQ-AAKVHIHISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyungmin Son et al.
Soft matter, 17(32), 7554-7564 (2021-08-03)
Materials that transform shapes responding to external stimuli can bring unprecedented innovations to soft matter physics, soft robotics, wearable electronics, and architecture. As most conventional soft actuation technologies induce large deformations only in a preprogrammed manner at designated locations, the
Z Rozynek et al.
Soft matter, 17(16), 4413-4425 (2021-04-29)
Electrically insulating objects immersed in a weakly conducting liquid may Quincke rotate when subjected to an electric field. Experimental and theoretical investigations of this type of electrorotation typically concern rigid particles and particle-free droplets. This work provides the basic features
Khobaib Khobaib et al.
Physical review. E, 103(6-1), 062605-062605 (2021-07-18)
Understanding of the behavior of an individual droplet suspended in a liquid and subjected to a stress is important for studying and designing more complex systems, such as emulsions. Here, we present an experimental study of the behavior of a
Z Rozynek et al.
Soft matter, 14(24), 5140-5149 (2018-06-09)
Droplets covered by adsorbed particles are used in a wide range of research studies and applications, including stabilising emulsions used in the food or cosmetic industries, and fabricating new materials, such as microcapsules or multi-cavity structures. Pickering emulsions are commonly
Zbigniew Rozynek et al.
ACS applied materials & interfaces, 11(25), 22840-22850 (2019-05-31)
Active, tunable, and reversible opening and closing of particle shells on droplets may facilitate chemical reactions in droplets and enable various small-scale laboratory operations, including online detection, measurement, and adjustment of droplet liquid. Manipulating various types of particle shells in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service