Skip to Content
Merck
All Photos(1)

Documents

78886

Millipore

Cetrimide Broth

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Pseudomonas Selective Broth, CB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein hydrolysate, 10 g/L
cetrimide, 0.2 g/L
gelatine peptone, pancreatic, 16 g/L
magnesium chloride, 1.4 g/L
potassium sulfate, 10 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
veterinary
water monitoring

microbiology

suitability

selective for Pseudomonas spp.

Application

For the isolation of Pseudomonas aeruginosa from food products.

Preparation Note

Dissolve 37.6 g in 950 ml distilled water, add 10 ml glycerol and sterilize by autoclaving at 121°C for 15 minutes. The prepared broth may contain slight precipitates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service