Skip to Content
Merck
All Photos(1)

Key Documents

74882

Supelco

Sodium 1-octanesulfonate monohydrate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

1-Octanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH2SO3Na · H2O
CAS Number:
Molecular Weight:
234.29
Beilstein:
3574241
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

sheet

quality

LiChropur

mol wt

234.29 g/mol

technique(s)

ion pair chromatography: suitable

mp

≥300 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

O.[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na.H2O/c1-2-3-4-5-6-7-8-12(9,10)11;;/h2-8H2,1H3,(H,9,10,11);;1H2/q;+1;/p-1

InChI key

MBURIAHQXJQKRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-octanesulfonate monohydrate is an ion-pair reagent suitable for cation separation sorted by carbon chain length.

Application

Sodium 1-octanesulfonate monohydrate may be used as an ion pair reagent for the determination of trigonelline in coffee samples by ion-pair chromatography. It may also be used as a mobile phase additive and diluent in the identification, assay and impurity profile analysis of methylseleno-L-cysteine (L-SeMC) bulk drug by reversed-phase ion-pairing HPLC method with UV detection.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and simple analysis of paraquat in tissue homogenate by ultra-high performance liquid chromatography.
Merritt, T. J. S., et al.
Analytical Methods : Advancing Methods and Applications, 3, 1428-1432 (2011)
Validation of a Stability-Indicating Method for Methylseleno-l-Cysteine (l-SeMC)
Canady K, et al.
Journal of Chromatographic Science, 54(1), 22-27 (2016)
Low pressure ion pair chromatography with amperometric detection for the determination of trigonelline in coffee samples
Sousa MBT, et al.
Food Research International, 114, 223-229 (2018)
Kamila Syslová et al.
Journal of chromatography. A, 1218(21), 3382-3391 (2011-03-15)
A sensitive assay method was developed for a parallel, rapid and precise determination of dopamine and its metabolites, homovanillic acid, 3-methoxytyramine and 3,4-dihydroxyphenylacetic acid, from brain microdialysates. The method consisted of a pre-treatment step, freeze-drying (lyophilization), to concentrate dopamine and
Maria Addolorata Saracino et al.
Journal of pharmaceutical and biomedical analysis, 104, 122-129 (2014-12-17)
A new microextraction by packed sorbent (MEPS) procedure coupled to a high-performance liquid chromatographic method has been developed for quantitation of catecholamines (i.e. norepinephrine, epinephrine and dopamine) in innovative biological samples, namely dried plasma and urine spots. Analyses were carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service