69964
MRS Agar
suitable for microbiology, NutriSelect® Basic
Synonym(s):
Lactobacillus Agar according to DeMan, Rogosa and Sharpe
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 12 g/L
dipotassium hydrogen phosphate, 2 g/L
D(+)-glucose, 20 g/L
magnesium sulfate (equivalent to 0.2 g/lheptahydrate), 0.1 g/L
manganous sulfate monhydrate, 0.05 g/L
meat extract, 5 g/L
sodium acetate, 5 g/L
triammonium citrate, 2 g/L
universal peptone, 10 g/L
yeast extract, 5 g/L
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbiological culture: suitable
final pH
6.5±0.2 (25 °C)
application(s)
agriculture
bioburden testing
environmental
food and beverages
microbiology
suitability
nonselective for Lactobacillus spp.
selective by low pH for Bifidobacterium spp.
Related Categories
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service