Skip to Content
Merck
All Photos(1)

Key Documents

69697

Supelco

1-Methylpyridinium iodide

analytical standard

Synonym(s):

Pyridine methiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8IN
CAS Number:
Molecular Weight:
221.04
Beilstein:
3707506
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (AT)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C[N+]1=CC=CC=C1.[I-]

InChI

1S/C6H8N.HI/c1-7-5-3-2-4-6-7;/h2-6H,1H3;1H/q+1;/p-1

InChI key

HLNJFEXZDGURGZ-UHFFFAOYSA-M

General description

1-Methylpyridinium iodide, a quaternary ammonium compound, is a well-known pyridinium salt. This six-membered ring heterocycle is generally found as a component of flavor and fragrances as well as food and cosmetic products.

Application

1-Methylpyridinium iodide may be used as an analytical standard for the determination of the analyte in food products by various analytical techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkylpyridiniums. 1. Formation in model systems via thermal degradation of trigonelline
Stadler RH, et al.
Journal of Agricultural and Food Chemistry, 50(5), 1192-1199 (2002)
HPLC analysis of aliphatic sulfonate surfactants using ion-pair detection
Zahrobsky M, et al.
Molecules (Basel), 10(9), 1179-1189 (2005)
The application of a new aromaticity index to six-membered ring heterocycles
Bird CW
Tetrahedron, 42(1), 89-92 (1986)
Zsigmond Benko et al.
Cell & bioscience, 7, 5-5 (2017-01-18)
HIV-1 protease (PR) is an essential enzyme for viral production. Thus, PR inhibitors (PIs) are the most effective class of anti-HIV drugs. However, the main challenge to the successful use of PI drugs in patient treatment is the emergence of
Identification of N-methyl-?-picolinium and other quaternary ammonium compounds from the oyster
Yasumoto T, et al.
Bulletin Of The Japanese Society Of Scientific Fisheries, 44(9), 529-529 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service