Skip to Content
Merck
All Photos(2)

Key Documents

68790

Supelco

Drometrizole trisiloxane

analytical standard

Synonym(s):

2-(2H-Benzotriazol-2-yl)-4-methyl-6-{2-methyl-3-{1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]disiloxanyl}propyl}phenol, Silatrizole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39N3O3Si3
CAS Number:
Molecular Weight:
501.84
Beilstein:
11343300
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(Cc1cc(C)cc(c1O)-n2nc3ccccc3n2)C[Si](C)(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C24H39N3O3Si3/c1-18-14-20(15-19(2)17-33(9,29-31(3,4)5)30-32(6,7)8)24(28)23(16-18)27-25-21-12-10-11-13-22(21)26-27/h10-14,16,19,28H,15,17H2,1-9H3

InChI key

HUVYTMDMDZRHBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Drometrizole trisiloxane (DRT) belongs to the lipophilic benzotriazole derivative class of compounds commonly utilized in sunscreen cosmetics.

Application

DRT may be used as a reference standard for the determination of DRT in:
  • Active pharmaceutical ingredients (APIs), formulations and stability samples by reversed phase ultra-high performance liquid chromatography (RP-UPLC) coupled to electrospray ionization-quadrupole time-of-flight tandem mass spectrometry (ESI-QTOF-MS/MS).
  • Suncare formulations by high performance liquid chromatography (HPLC) with ultraviolet (UV) detection.
  • Cosmetic products by LC-UV.
  • Sewage sludge samples by QuEChERS (quick, easy, cheap, effective, rugged, and safe) extraction procedure and gas chromatography combined with MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sensitive, Stability indicating UPLC method for the identification and characterization of forced degradation products for Drometrizole Trisiloxane through MSn studies.
Babu MA, et al.
Journal of Applied Pharmaceutical Science, 8(06), 065-074 (2018)
A reliable and environmentally-friendly liquid-chromatographic method for multi-class determination of fat-soluble UV filters in cosmetic products.
Chisvert A, et al.
Analytica Chimica Acta, 790(1-2), 61-67 (2013)
Development and optimization of a QuEChERS-GC-MS/MS methodology to analyse ultraviolet-filters and synthetic musks in sewage sludge.
Ramos S, et al.
The Science of the Total Environment, 790(1-2), 61-67 (2018)
Determination of sixteen UV filters in suncare formulations by high-performance liquid chromatography.
Schakel DJ, et al.
Journal of Chromatography A, 1049(1-2), 127-130 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service