Skip to Content
Merck
All Photos(2)

Key Documents

53882

Supelco

Disperse Orange 3

analytical standard

Synonym(s):

4-(4-Nitrophenylazo)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC6H4NH2
CAS Number:
Molecular Weight:
242.23
Colour Index Number:
11005
Beilstein:
1842907
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~200 °C (dec.) (lit.)

λmax

443 nm

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H10N4O2/c13-9-1-3-10(4-2-9)14-15-11-5-7-12(8-6-11)16(17)18/h1-8H,13H2/b15-14+

InChI key

UNBOSJFEZZJZLR-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Orange 3 is widely used in textile industry for dyeing polyester fibers.

Application

Disperse Orange 3 may be used as a test material for investigating its degradation by the white rot fungus Pleurotus ostreatus and the products obtained upon degradation are determined using UV-visible spectrophotometric and high-performance liquid chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC and spectrophotometric analysis of biodegradation of azo dyes by Pleurotus ostreatus
Zhao X and Hardin RI
Dyes and Pigments, 73(3), 322-325 (2007)
A A Ahmad et al.
Journal of hazardous materials, 170(2-3), 612-619 (2009-06-12)
The purpose of this work is to obtain optimal preparation conditions for activated carbons prepared from rattan sawdust (RSAC) for removal of disperse dye from aqueous solution. The RSAC was prepared by chemical activation with phosphoric acid using response surface
Laura Malinauskiene et al.
Contact dermatitis, 68(2), 65-75 (2013-01-08)
Several disperse dyes (DDs) are still considered to be the most important allergens in textile dermatitis, but there are sparse data about their current use in textiles. The aim of this review was to evaluate published studies and reports on
Manfred Kneilling et al.
Experimental dermatology, 19(5), 435-441 (2010-02-02)
Patients sensitized to para-phenylenediamine (PPD) by semi-permanent tattoos increasingly develop threatening allergic reactions in response to black hair dye. The gold standard to diagnose allergic contact dermatitis is to perform epicutaneous patch tests, however, iatrogenic sensitizations and severe patch test
C Berger et al.
Contact dermatitis, 10(3), 154-157 (1984-03-01)
Thin-layer (uni-dimensional and bi-dimensional) chromatography studies were performed on 52 beige stockings and pantyhose from different countries. They demonstrated the presence of Disperse Yellow 3 in 51 and Disperse Orange 3 in 15 stockings. In the absence of isolation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service