Skip to Content
Merck
All Photos(1)

Key Documents

51759

Millipore

Rapid Enterococci ChromoSelect Agar

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Enterococci Rapid ChromoSelect Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
chromogenic mixture, 0.06 g/L
disodium hydrogen phosphate, 1.25 g/L
peptone special, 10 g/L
polysorbate 80, 2 g/L
sodium azide, 0.3 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Plus

impurities

sodium azide

final pH

7.5±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-8°C

suitability

selective and differential for Enterococcus spp.
selective and differential for Staphylococcus spp.

Application

Rapid Enterococci ChromoSelect Agar is used for rapid and easy identification and differentiation of Enterococci from water samples. It contains chromogenic substrate, which aids in the detection of Enterococci from water samples.

Preparation Note

Suspend 33.6 g in 1 litre distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 1 bar pressure (121°C) for 15 minutes. Cool to 50 °C and pour into sterile petri plates.

Other Notes

Medium for the assay of water for Enterococci

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Manafi. R. Sommer
Water Science and Technology, 27, 271-271 (1993)
H Althaus et al.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale A, Medizinische Mikrobiologie, Infektionskrankheiten und Parasitologie = International journal of microbiology and hygiene. A, Medical microbiology, infectious..., 252(2), 154-165 (1982-06-01)
The rules of the European Economic Community to harmonize the hygienic quality of drinking water write out the faecal streptococci as indicator organisms. This operating definition of the faecal streptococci prohibits its taxonomic limitation to enterococci or group D streptococci.
A new medium for the detection of enterococci in water.
W LITSKY et al.
American journal of public health and the nation's health, 43(7), 873-879 (1953-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service