Skip to Content
Merck
All Photos(1)

Key Documents

51707

Sigma-Aldrich

Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin

≥98.0%

Synonym(s):

TM-β-CD, TM‐β‐CyD, TRIMEB, 2,3,6-Tri-O-methyl-β-cyclodextrin, Trimethyl-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H112O35
CAS Number:
Molecular Weight:
1429.54
Beilstein:
78748
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (TLC)
≥98.0%

form

powder or crystals

optical activity

[α]25/D +157±4°, c = 1% in H2O

mp

170-178 °C (lit.)

solubility

H2O: 50 mg/mL, slightly turbid, colorless

functional group

ether

storage temp.

2-8°C

SMILES string

COC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC)O[C@H](O[C@@H]4[C@@H](COC)O[C@H](O[C@@H]5[C@@H](COC)O[C@H](O[C@@H]6[C@@H](COC)O[C@H](O[C@@H]7[C@@H](COC)O[C@H](O[C@@H]8[C@@H](COC)O[C@H](O[C@H]1[C@H](OC)[C@H]2OC)[C@H](OC)[C@H]8OC)[C@H](OC)[C@H]7OC)[C@H](OC)[C@H]6OC)[C@H](OC)[C@H]5OC)[C@H](OC)[C@H]4OC)[C@H](OC)[C@H]3OC

InChI

1S/C63H112O35/c1-64-22-29-36-43(71-8)50(78-15)57(85-29)93-37-30(23-65-2)87-59(52(80-17)44(37)72-9)95-39-32(25-67-4)89-61(54(82-19)46(39)74-11)97-41-34(27-69-6)91-63(56(84-21)48(41)76-13)98-42-35(28-70-7)90-62(55(83-20)49(42)77-14)96-40-33(26-68-5)88-60(53(81-18)47(40)75-12)94-38-31(24-66-3)86-58(92-36)51(79-16)45(38)73-10/h29-63H,22-28H2,1-21H3/t29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43+,44+,45+,46+,47+,48+,49+,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-/m1/s1

InChI key

DSDAICPXUXPBCC-MWDJDSKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin is a β-cyclodextrin derivative that finds potential use in pharmaceutical, food and cosmetic applications. Its attractive features include: the non-polar cavity desirable for its use as a molecular host for a wide range of molecules and its affinity for organic compounds in solution as well as crystalline phases.[1]

Application

Chiral additive in liquid chromatography for enantiomer resolution. Chiral separation of basic drug substances by capillary electrophoresis.
Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin may be used:
  • To investigate the crystal structure of its complexes with m-iodophenol, 4-biphenylacetic acid[2] and (R)- and (S)-flurbiprofen[3] by X-ray analysis.
  • To study the candidature of its complex with vitamin A for potential application as a drug delivery system for ophthalmic applications by high sensitivity fluorescence spectrometry and high pressure liquid chromatography (HPLC).[4]
  • In the determination of analyte composition in commercial samples by HPLC coupled to mass spectrometry (MS).[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structures of heptakis (2, 3, 6-tri-O-methyl)-beta-cyclodextrin complexes with (R)-and (S)-Flurbiprofen
Harata K, et al.
Journal of inclusion phenomena, 6(5), 443-460 (1988)
Hiroko F Kasai et al.
Chemical & pharmaceutical bulletin, 53(10), 1270-1276 (2005-10-06)
Separation of the enantiomers of each of three different racemates, neutral rac-alpha-tetralol, acidic rac-2-phenylpropionic acid, and basic rac-1-phenylethylamine, using subcritical and supercritical fluid chromatography with two different chiral stationary phases, heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin (Sumichiral OA-7500 column) and tris(3,5-dimethylphenylcarbamate) of amylose (Chiralpak AD-H
Alessandra Ammazzalorso et al.
Journal of chromatography. A, 1088(1-2), 110-120 (2005-09-01)
The enantiomeric separation of gemfibrozil chiral analogues was performed by capillary zone electrophoresis (CZE). Resolution of the enantiomers was achieved using heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin (TM-beta-CD) as chiral selector dissolved into a buffer solution. In order to optimize the separation conditions, type, pH
Stabilization of all-trans-retinol by cyclodextrins: a comparative study using HPLC and fluorescence spectroscopy
Semenova EM, et al.
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 44(1-4), 155-158 (2002)
Franca Castiglione et al.
The journal of physical chemistry. B, 115(29), 9005-9013 (2011-06-15)
The triblock-copolymer poly(ethylene oxide)-poly(propyleneoxide)-poly(ethylene oxide) (PEO-PPO-PEO), referred to as Pluronic, is widely studied for its unique aggregation properties and its applications in drug delivery and targeting. In previous studies [Dreiss, C. A.; et al. Soft Matter 2009, 5, 1888-1896], we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service