Skip to Content
Merck
All Photos(1)

Key Documents

50875

Millipore

GSP Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Pseudomonas Aeromonas Selective Agar acc. to Kielwein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
magnesium sulfate, 0.5 g/L
phenol red, 0.36 g/L
potassium dihydrogen phosphate, 2 g/L
sodium L-glutamate, 10 g/L
starch (soluble), 20 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

Aeromonas spp.
selective and differential for Pseudomonas spp.

General description

Agar is a complex gelatinous carbohydrate, and is added to the LB broth (Lysogeny broth, Luria broth, Lennox broth, or Luria-Bertani medium), in order to form gel for bacteria to grow upon. LB Broth is the most widely used nutritionally rich medium, for the culture and growth of bacteria. There are several different formulations of Lb broth, but the composition is generally the same comprising of peptides and casein peptones, trace elements, mineral and vitamins.

Application

For the detection of Pseudomonas and Aeromonas in all types of food as well as in the equipment and water supply of food-processing firms.

Preparation Note

Dissolve 45 g in 1 litre distilled water and sterilize by autoclaving at 121°C for 15 minutes. Cool to 45-50°C and add 100′000 IU/l Penicillin G (Cat. No. 13752; 60mg/l) and if desired 10 mg/l Pimaricin (Cat. No. 80482; 400ul/l). Mix well and pour into sterile petri plates.

Other Notes

A medium for selective cultivation of Pseudomonas and Aeromonas; The aerobic Pseudomonas - a taxonomic study

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.Y. Stanier et al.
Journal of General Microbiology, 42, 159-159 (1966)
G. Kielwein
Archiv Fur Lebensmittelhygiene, 20, 131-131 (1969)
Archiv Fur Lebensmittelhygiene, 22, 15-15 (1971)
The structure of agar: Part I. Fractionation of a complex mixture of polysaccharides.
Duckworth M and Yaphe W.
Carbohydrate Research, 16 (1), 189-197 (1971)
Agar and agarose biotechnological applications.
Armisen R.
Hydrobiologia, 221 (1), 157-166 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service