Skip to Content
Merck
All Photos(1)

Key Documents

50484

Sigma-Aldrich

Methylene Blue solution

suitable for microbiology

Synonym(s):

Ehrlich’s reagent III, Loeffler’s Methylene Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Beilstein:
3641570
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

shelf life

limited shelf life, expiry date on the label

concentration

0.5% in H2O (0.5 g/100 mL)

technique(s)

microbe id | staining: suitable

application(s)

microbiology

suitability

Bacillus spp.
Corynebacterium spp.
bacteria

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methylene Blue was earlier recognized as a good nuclear stain, the staining solutions are always either aged or alkalized, if not both. More importantly it is an excellent stain for ribonucleic acid, which is the important stainable constituent of bacterial bodies, of lymphocyte and hemoportoal cytoplasm, and of nerve cell tigroid granules. It can also be employed in capsule stain. The capsule stain begins as a negative stain leaving the organisms colorless. The capsule are clearly seen as halo surrounding the bacterial cell. Another staining procedure is performed with nigrosin (Cat. No. 43925).

Components

reagent contains:
methylene blue 0.5 g
distilled water 100 ml

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel Gehrung et al.
Journal of biomedical optics, 24(12), 1-11 (2019-10-19)
Photoacoustic tomography (PAT) is intrinsically sensitive to blood oxygen saturation (sO2) in vivo. However, making accurate sO2 measurements without knowledge of tissue- and instrumentation-related correction factors is extremely challenging. We have developed a low-cost flow phantom to facilitate validation of
Q R J G Tummers et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 40(7), 850-858 (2014-05-28)
Despite recent developments in preoperative breast cancer imaging, intraoperative localization of tumor tissue can be challenging, resulting in tumor-positive resection margins during breast conserving surgery. Based on certain physicochemical similarities between Technetium((99m)Tc)-sestamibi (MIBI), an SPECT radiodiagnostic with a sensitivity of
Jonel Steffen et al.
The British journal of ophthalmology, 98(1), 13-15 (2013-10-26)
To evaluate the diagnostic accuracy of methylene blue used as a non-invasive in vivo stain to detect ocular surface squamous neoplasia (OSSN). A test validation study was performed according to Standards for the reporting of diagnostic accuracy studies (STARD) guidelines
Muhannad N Fadhel et al.
Journal of biomedical optics, 24(11), 1-8 (2019-11-11)
Solid tumors are typically supplied nutrients by a network of irregular blood vessels. By targeting these vascular networks, it might be possible to hinder cancer growth and metastasis. Vascular disrupting agents induce intertumoral hemorrhaging, making photoacoustic (PA) imaging well positioned
Bram Vivijs et al.
Applied and environmental microbiology, 80(19), 6054-6061 (2014-07-27)
Some members of the family Enterobacteriaceae ferment sugars via the mixed-acid fermentation pathway. This yields large amounts of acids, causing strong and sometimes even lethal acidification of the environment. Other family members employ the 2,3-butanediol fermentation pathway, which generates comparatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service