Expiration dated products are not retested by the supplier, and expiration dates are not extended. The recommended retest date is not printed on container labels.
49377-U
ProClin™ 150
ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 1.5%, pkg of 400 mL
Synonym(s):
1.5% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one
About This Item
Recommended Products
Quality Level
description
biocide for IVD use
form
liquid
shelf life
2 yr
contains
23 to 25% Mg salts
composition
CMIT/MIT, 1.5%
packaging
pkg of 400 mL
parameter
operating temperature at <45 °C
matrix
Water
operating pH range
2.5-8.5
pH range
1.7-3.7
bp
100 °C
mp
-21 °C
solubility
water: soluble
density
1.20 g/cm3
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Would it be possible to grant an expiration date extension of a minimum of 6 months for the lot that has expired?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service