46022
Pentyl acetate
puriss. p.a., ≥98.5% (GC)
Synonym(s):
Amyl acetate
About This Item
Recommended Products
vapor density
4.5 (vs air)
Quality Level
vapor pressure
4 mmHg ( 20 °C)
grade
puriss. p.a.
Assay
≥98.5% (GC)
autoignition temp.
680 °F
impurities
≤1% isopentyl acetate
refractive index
n20/D 1.402 (lit.)
n20/D 1.403
bp
142-149 °C (lit.)
mp
−100 °C (lit.)
density
0.876 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
functional group
ester
SMILES string
CCCCCOC(C)=O
InChI
1S/C7H14O2/c1-3-4-5-6-9-7(2)8/h3-6H2,1-2H3
InChI key
PGMYKACGEOXYJE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
105.8 °F
Flash Point(C)
41 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service