Skip to Content
Merck
All Photos(1)

Documents

46003

Supelco

β-Cyfluthrin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18Cl2FNO3
CAS Number:
Molecular Weight:
434.29
Beilstein:
2788149
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2ccc(F)c(Oc3ccccc3)c2

InChI

1S/C22H18Cl2FNO3/c1-22(2)15(11-19(23)24)20(22)21(27)29-18(12-26)13-8-9-16(25)17(10-13)28-14-6-4-3-5-7-14/h3-11,15,18,20H,1-2H3

InChI key

QQODLKZGRKWIFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos G Athanassiou et al.
Journal of economic entomology, 106(2), 1064-1070 (2013-06-22)
The insecticidal effect of Temprid, a formulation that contains beta-cyfluthrin and imidacloprid, was tested on concrete for control of seven stored-product insect species: the rusty grain beetle, Cryptolestes ferrugineus (Stephens); the sawtoothed grain beetle, Oryzaephilus surinamensis (L.); the red flour
Changlu Wang et al.
Journal of economic entomology, 106(1), 347-352 (2013-03-02)
Current bed bug, Cimex lectularius L., control usually involves insecticide applications that pose a high risk of insecticide exposure to residents and applicators. To minimize these risks and the amount of insecticides used, we designed and evaluated a dust band
Sandra A Allan
Journal of vector ecology : journal of the Society for Vector Ecology, 36(1), 59-67 (2011-06-04)
Mosquitoes characteristically feed on plant-derived carbohydrates and honeydew just after emergence and intermittently during their lives. Development of toxic baits focusing on this carbohydrate-seeking behavior may potentially contribute to localized control. In the present study, ten insecticides were fed to
Kumelachew Mulu Loha et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(7), 687-691 (2012-05-09)
In the present investigation, bioefficacy of developed β-cyfluthrin formulations, utilizing laboratory synthesized poly(ethylene glycols) based amphiphilic copolymers, were evaluated against Callosobruchus maculatus (Coleoptera: Bruchidae). The bioefficacy data indicated that the formulations developed by utilizing polymers having PEG - 1500 (3c)
Shaohua Chen et al.
Bioresource technology, 132, 16-23 (2013-02-12)
Brevibacterium aureum DG-12, a new bacterial strain isolated from active sludge, was able to degrade and utilize cyfluthrin as a growth substrate in the mineral medium. Response surface methodology using central composite rotatable design of cultural conditions was successfully employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service