Skip to Content
Merck
All Photos(1)

Documents

45883

Supelco

Metolachlor solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Beilstein:
8396147
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CCc1cccc(C)c1N(C(C)COC)C(=O)CCl

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3

InChI key

WVQBLGZPHOPPFO-UHFFFAOYSA-N

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Restivo et al.
Journal of hazardous materials, 239-240, 249-256 (2012-09-27)
The catalytic ozonation of the herbicide metolachlor (MTLC) was tested using carbon nanomaterials as catalysts. Multiwalled carbon nanotubes were used in semi-batch experiments and carbon nanofibres grown on a honeycomb cordierite monolith were tested in continuous experiments. The application of
Huong Mai et al.
Marine pollution bulletin, 64(12), 2663-2670 (2012-11-06)
This study evaluated embryotoxicity and genotoxicity of two dissolved metals copper and cadmium (Cu and Cd) and two pesticides (metolachlor and irgarol) occurring in Arcachon Bay (SW France) in Pacific oyster (Crassostrea gigas) larvae and investigated the relationship between those
M T Moore et al.
Bulletin of environmental contamination and toxicology, 89(2), 292-295 (2012-06-02)
Phytotoxicity assessments were performed to compare responses of Typha latifolia (L.) seeds to atrazine (only) and atrazine + S-metolachlor exposure concentrations of 0.03, 0.3, 3, and 30 mg L(-1), as well as permethrin exposure concentrations of 0.008, 0.08, 0.8, and
Megha Thakkar et al.
Aquatic toxicology (Amsterdam, Netherlands), 126, 198-206 (2012-12-12)
Metolachlor, a chloroacetanilide herbicide, has been frequently detected in coastal waters. This study examined the growth, photosynthesis, and detoxification responses of chlorophyte Dunaliella tertiolecta (DT) and brown tide alga Aureococcus anophagefferens (AA) upon 5-day exposure to 0.5-5 mg L(-1) metolachlor.
Zisis Vryzas et al.
Chemosphere, 89(11), 1330-1338 (2012-06-29)
Biotransformation studies of atrazine, metolachlor and evolution of their metabolites were carried out in soils and subsoils of Northern Greece. Trace atrazine, its metabolites and metolachlor residues were detected in field soil samples 1 year after their application. The biotransformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service