Skip to Content
Merck
All Photos(1)

Key Documents

45674

Sigma-Aldrich

Erythromycin

tested according to Ph. Eur.

Synonym(s):

Erythromycinum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
Beilstein:
75279
EC Number:
MDL number:
UNSPSC Code:
51282304
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Agency

EPA 1694
USP/NF
tested according to Ph. Eur.

Quality Level

form

solid

color

white to faint yellow

solubility

H2O: soluble 2 mg/mL
acetone: freely soluble
acetonitrile: freely soluble
alcohol: soluble
amyl acetate: moderately soluble
chloroform: soluble
diethyl ether: soluble
ethyl acetate: freely soluble

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

environmental

Mode of action

protein synthesis | interferes

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

ULGZDMOVFRHVEP-RWJQBGPGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Erythromycin is an antibiotic produced by growth of certain strains of Streptomyces erythreus. This product is composed largely of erythromycin A with small amounts of erythromycins B and C and is recommended for concentration at 100 mg/L. Concentrations between 50 and 200 mg/L have also proven effective in controlling bacterial growth. Erythromycin has been used as a motilin receptor agonist, to block respiratory glycoconjugate secretion in human airways in vitro††, and for selecting plasmid-cured and recombinant lactococcus lactis MG1363 strains.

Biochem/physiol Actions

Mode of Action: Erythromycin acts by inhibiting elongation at the transpeptidation step, specifically aminoacyl translocation from the A-site to P-site by binding to the 50s subunit of the bacterial 70s rRNA complex.

Antimicrobial Spectrum: This product acts against both gram-negative and gram-positive bacteria.

Packaging

25g

Caution

This product is stable in solution at 37°C for 3 days. Stock solutions should be stored at 2-8°C.

Preparation Note

This product is soluble in water at 2 mg/mL, with a 0.067% solution in water yielding a pH of 8.0-10.5. It is also soluble in ethanol at 50 mg/mL, yielding a clear, colorless to faint yellow solution. It is freely soluble in alcohol, acetone, chloroform, acetonitrile and ethyl acetate but forms salts with acids. All solutions should be protected from light.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Peeters et al.
The American journal of physiology, 257(3 Pt 1), G470-G474 (1989-09-01)
Erythromycin A (EMA) is a potent stimulator of gastrointestinal motor activity. In vitro studies suggest that it mimics motilin, a peptide that stimulates motor activity in human and in rabbit via smooth muscle receptors. We have compared the in vitro
Elisabet I Nielsen et al.
Antimicrobial agents and chemotherapy, 51(1), 128-136 (2006-10-25)
Dosing of antibacterial agents is generally based on point estimates of the effect, even though bacteria exposed to antibiotics show complex kinetic behaviors. The use of the whole time course of the observed effects would be more advantageous. The aim
Anna C Shore et al.
Antimicrobial agents and chemotherapy, 54(12), 4978-4984 (2010-10-06)
The staphylococcal cfr gene mediates resistance to phenicols, lincosamides, oxazolidinones, pleuromutilins, and streptogramin A, a phenotype that has been termed PhLOPS(A). The cfr gene has mainly been associated with coagulase-negative staphylococcal isolates from animals, and only a few cfr-positive methicillin-resistant
Leena Keurulainen et al.
Journal of medicinal chemistry, 53(21), 7664-7674 (2010-10-12)
Chlamydia pneumoniae is an intracellular bacterium that responds poorly to antibiotic treatment. Insufficient antibiotic usage leads to chronic infection, which is linked to disease processes of asthma, atherosclerosis, and Alzheimer's disease. The Chlamydia research lacks genetic tools exploited by other
Nicole Wolter et al.
Antimicrobial agents and chemotherapy, 51(3), 1092-1095 (2007-01-11)
A rare clinical isolate of Streptococcus pneumoniae, highly resistant to telithromycin, contained erm(B) with a truncated leader peptide and a mutant ribosomal protein L4. By transformation of susceptible strains, this study shows that high-level telithromycin resistance is conferred by erm(B)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service