Skip to Content
Merck
All Photos(1)

Key Documents

45644

Supelco

Propoxur

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO3
CAS Number:
Molecular Weight:
209.24
Beilstein:
1879891
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1ccccc1OC(C)C

InChI

1S/C11H15NO3/c1-8(2)14-9-6-4-5-7-10(9)15-11(13)12-3/h4-8H,1-3H3,(H,12,13)

InChI key

ISRUGXGCCGIOQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propoxur is a non-systemic carbamate insecticide, mainly used for agricultural and non-agricultural applications, for controlling a variety of insects pests such as ants, crickets flies, mosquitoes, cockroaches, chewing and sucking insects.

Application

Propoxur may be used as an analytical reference standard for the quantification of the analyte in environmental samples, beverages and vegetable samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MSPD-GC-MS-MS determination of residues of 15 organic nitrogen-containing pesticides in vegetables
Chromatographia, 72(11-12), 1143-1148 (2010)
Amine Gizem Canlı et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-22)
A sensitive, rapid, reliable, and easily applicable method based on magnetic solid phase extraction (MSPE) combined with HPLC-PDA was developed for monitoring propoxur (PRO) and fenitrothion (FEN) pesticides in environmental water samples. The effect of major experimental variables on the
Finding of pesticides in fashionable fruit juices by LC-MS/MS and GC-MS/MS
Wu J, et al.
Microchimica Acta, 166(1-2), 157-162 (2009)
Enrique M Ostrea et al.
Neurotoxicology, 33(4), 669-675 (2011-12-14)
Our aim was to determine the effects of fetal exposure to propoxur and pyrethroids, on child neurodevelopment at 2 years of age. Mothers were prospectively recruited during mid-pregnancy in Bulacan, Philippines where multiple pesticides including propoxur, cyfluthrin, chlorpyrifos, cypermethrin, pretilachlor
Mo'awia Mukhtar Hassan et al.
Parasites & vectors, 5, 46-46 (2012-03-09)
Phlebotomus papatasi the vector of cutaneous leishmaniasis (CL) is the most widely spread sand fly in Sudan. No data has previously been collected on insecticide susceptibility and/or resistance of this vector, and a first study to establish a baseline data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service