Skip to Content
Merck
All Photos(1)

Key Documents

45464

Supelco

DNOC

PESTANAL®, analytical standard

Synonym(s):

2-Methyl-4,6-dinitrophenol, 3,5-Dinitro-2-hydroxytoluene, 4,6-Dinitro-2-methylphenol, 4,6-Dinitro-o-cresol, DNOC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2O5
CAS Number:
Molecular Weight:
198.13
Beilstein:
2054389
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10H,1H3

InChI key

ZXVONLUNISGICL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Ye et al.
Water research, 43(5), 1303-1312 (2009-01-27)
The adsorption and degradation of 4,6-o-dinitrocresol (DNOC) and p-nitrophenol (PNP) in SWy-2 montmorillonite clay slurry were investigated. The pH and type of cation of the slurry were varied. Results showed that adsorption of DNOC and PNP increased at lower pH
Fanny Marlin et al.
Chembiochem : a European journal of chemical biology, 13(17), 2593-2598 (2012-11-07)
Oligonucleotides and their analogues, such as peptide nucleic acids (PNAs), can be used in chemical strategies to artificially control gene expression. Inefficient cellular uptake and inappropriate cellular localization still remain obstacles in biological applications, however, especially for PNAs. Here we
Hanh Nguyen-Ngoc et al.
Ecotoxicology and environmental safety, 72(2), 316-320 (2008-06-17)
Synchronous-scan spectrofluorometry was applied to Chlorella vulgaris cells to assess the toxicity of heavy metals and herbicides in water. Simultaneous scan of both the excitation and emission spectra was done at a constant wavelength difference Deltalambda (20-140 nm) between the
Nicholas J Kotloski et al.
mBio, 4(1), doi:10-doi:10 (2013-01-17)
Shewanella oneidensis strain MR-1 is widely studied for its ability to respire a diverse array of soluble and insoluble electron acceptors. The ability to breathe insoluble substrates is defined as extracellular electron transfer and can occur via direct contact or
Jolanta Bieganska
Environmental science & technology, 39(4), 1190-1196 (2005-03-19)
Neutralization of 4,6-dinitro-o-cresol, of which the best-before date had expired and which could not be used any longer, was carried out by means of the detonative combustion method. The pesticide comprises two nitro groups and is called DNOC. Composition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service