Skip to Content
Merck
All Photos(1)

Key Documents

45404

Supelco

Coumatetralyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O3
CAS Number:
Molecular Weight:
292.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC1=C(C2CCCc3ccccc23)C(=O)Oc4ccccc14

InChI

1S/C19H16O3/c20-18-15-9-3-4-11-16(15)22-19(21)17(18)14-10-5-7-12-6-1-2-8-13(12)14/h1-4,6,8-9,11,14,20H,5,7,10H2

InChI key

ULSLJYXHZDTLQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coumatetralyl is classified under the hydroxycoumarin group of anticoagulant rodenticides.

Application

Coumatetralyl may be used as an analytical reference standard for the determination of coumatetralyl in:
  • Sludge samples by solid-liquid extraction (SLE) and liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) equipped with electrospray ionization (ESI), and selected reaction monitoring (SRM) detection.
  • Wastewater samples by miniaturized liquid-liquid extraction (LLE), solid-phase extraction (SPE) and LC-ESI-MS/MS with SRM detection.
  • Vertebrate wildlife and domestic animals by gel permeation chromatography (GPC) clean-up followed by ultra-high performance liquid chromatography (UHPLC) coupled to ESI-MS/MS with multiple reaction monitoring (MRM) detection.
  • Blood samples by LLE and ESI-LC-MS/MS with MRM detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

A validated LC-MS-MS method for simultaneous identification and quantitation of rodenticides in blood.
Bidny S, et al.
Journal of Analytical Toxicology, 39(3), 219-224 (2015)
G Chopra et al.
The Journal of hygiene, 94(3), 327-330 (1985-06-01)
Coumatetralyl was evaluated in the laboratory against Bandicota bengalensis and Rattus rattus. Feeding tests with 0.0375% coumatetralyl baits produced 100% mortality after a single day feeding period in B. bengalensis and after a 10-day period in R. rattus. The anticoagulant
Comprehensive characterization of anticoagulant rodenticides in sludge by liquid chromatography-tandem mass spectrometry.
Gomez-Canela C and Lacorte S
Environmental Science and Pollution Research International, 23(15), 15739-15748 (2016)
Comprehensive characterization of rodenticides in wastewater by liquid chromatography-tandem mass spectrometry.
Gomez-Canela C, et al.
Analytical and Bioanalytical Chemistry, 406(1), 345-358 (2014)
Mi-cong Jin et al.
Journal of applied toxicology : JAT, 28(5), 621-627 (2007-11-03)
A high-performance liquid chromatographic-tandem mass spectrometric (HPLC-MS-MS) method was developed and validated to determine simultaneously coumafuryl and coumateralyl in animal tissues using warfarin as an internal standard (IS). Animal tissue samples were extracted with ethyl acetate and cleaned by Oasis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service