Skip to Content
Merck
All Photos(1)

Documents

45345

Sigma-Aldrich

Epoxy embedding medium

for microscopy

Synonym(s):

Epon 812 substitute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102904
NACRES:
MA.02

biological source

synthetic

Quality Level

grade

for microscopy

form

viscous liquid

impurities

glycerol glycidyl ethers

color

colorless to faint yellow

refractive index

n/D 1.480-1.483

suitability

suitable for (use as an embedding medium)

application(s)

hematology
histology

storage temp.

room temp

General description

additional information see 45359 Epoxy Embedding Medium kit

Other Notes

Embedding media for electron microscopy

Legal Information

Epon is a trademark of Hexion, Inc.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exact anhydride epoxy percentages for electron microscopy embedding (Rpon).
C N Burke et al.
Journal of ultrastructure research, 36(1), 119-126 (1971-07-01)
Otilia Zarnescu et al.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada, 23(3), 668-678 (2017-05-11)
Cytological responses in different organs of sentinel organisms have proven to be useful tools for characterizing the health status of those organisms and assessing the impact of environmental contaminants. Our study shows that nickel (II) accumulated in both germ cells
Jingyuan Wang et al.
International journal of molecular medicine, 45(6), 1875-1887 (2020-04-03)
Multiple mechanisms are involved in regulating hepatic ischemia‑reperfusion injury (IRI), in which Kupffer cells (KCs), which are liver‑resident macrophages, play critical roles by regulating inflammation and the immune response. Suberoylanilide hydroxamic acid (SAHA), a pan‑histone deacetylase inhibitor, has anti‑inflammatory effects
T Martinovic et al.
Tissue & cell, 52, 92-100 (2018-06-03)
Type 2 diabetes is an endocrine disorder characterized with hyperglycemia, hyperinsulinemia and insulin resistance. Morphological changes in cell nuclei in diabetes were recently detected. The aim of this study was to compare electron microscopic features of lymphocyte nuclei in type
Tatyana Shulyatnikova et al.
Brain research, 1751, 147174-147174 (2020-11-12)
Endogenous toxicity caused by systemic inflammation as well as by acute liver failure triggers a wide range of dysfunctional disorders in the brain ranging from delirium and acute psychosis to coma. Astrocytes, the main homeostatic cells of the central nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service