Skip to Content
Merck
All Photos(1)

Key Documents

44941

Supelco

Iron(III) citrate tribasic monohydrate

18-20% Fe basis (T)

Synonym(s):

Ferric citrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5FeO7 · H2O
CAS Number:
Molecular Weight:
262.96
Beilstein:
4052503
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

form

solid

concentration

18-20% Fe (T)

anion traces

chloride (Cl-): ≤500 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
K: ≤500 mg/kg
Na: ≤200 mg/kg
Ni: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

functional group

hydroxyl

SMILES string

OC12CC(=O)O[Fe](OC(=O)C1)OC2=O

InChI

1S/C6H8O7.Fe/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+3/p-3

InChI key

NPFOYSMITVOQOS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Iron(III) citrate tribasic monohydrate may be used as the source of iron, in one of the compositions, used during the preparation of perfusion solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Gómez-Ayala et al.
Laboratory animals, 32(1), 72-79 (1998-03-03)
The effects of iron deficiency on the absorption of different dietary sources of iron were studied, together with the interactions between iron, calcium, phosphorus, magnesium, copper and zinc in the jejunum-ileum of control and iron-deficient rats. In this study, three
Tatsuya Fukushima et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(42), 16829-16834 (2012-10-03)
Citrate is a common biomolecule that chelates Fe(III). Many bacteria and plants use ferric citrate to fulfill their nutritional requirement for iron. Only the Escherichia coli ferric citrate outer-membrane transport protein FecA has been characterized; little is known about other
Marvin Sinsakul et al.
Nephron. Clinical practice, 121(1-2), c25-c29 (2012-10-19)
A phase II open-label study was conducted in hemodialysis patients evaluating the short-term safety, tolerability, and iron absorption with ferric citrate when used as a phosphate binder. Enrollment occurred in two periods. Period 1 recruited patients taking 6-15 pills/day of
Petra Vukosav et al.
Analytica chimica acta, 745, 85-91 (2012-09-04)
A detailed study of iron (III)-citrate speciation in aqueous solution (θ=25°C, I(c)=0.7 mol L(-1)) was carried out by voltammetric and UV-vis spectrophotometric measurements and the obtained data were used for reconciled characterization of iron (III)-citrate complexes. Four different redox processes
Jessica A Smith et al.
Applied and environmental microbiology, 79(3), 901-907 (2012-11-28)
Geobacter species are important Fe(III) reducers in a diversity of soils and sediments. Mechanisms for Fe(III) oxide reduction have been studied in detail in Geobacter sulfurreducens, but a number of the most thoroughly studied outer surface components of G. sulfurreducens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service