Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
2-(HO2C)C6H4CO2CH3
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
224-476-6
Beilstein/REAXYS Number:
2048766
MDL number:
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
mp
82-84 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
SMILES string
COC(=O)c1ccccc1C(O)=O
InChI
1S/C9H8O4/c1-13-9(12)7-5-3-2-4-6(7)8(10)11/h2-5H,1H3,(H,10,11)
InChI key
FNJSWIPFHMKRAT-UHFFFAOYSA-N
General description
mono-Methyl phthalate belongs to the class of phthalates, broadly employed as plasticizers in various domestic and industrial products, personal care products, pharmaceuticals, medical devices, and paints.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
mono-Methyl phthalate may be used as an analytical reference standard for the quantification of the analyte in biological samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) and gas chromatography-mass spectrometry.
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Still not finding the right product?
Explore all of our products under mono-Methyl phthalate
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phthalate metabolites in urine samples from school children in Taipei, Taiwan
Bao J, et al.
Archives of Environmental Contamination and Toxicology, 69(2), 202-207 (2015)
Simultaneous determination of multiple phthalate metabolites and bisphenol-A in human urine by liquid chromatography?tandem mass spectrometry
Chen L-M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 904(2), 73-80 (2012)
Da-Wei Liang et al.
Applied microbiology and biotechnology, 74(1), 221-229 (2006-11-11)
Results of batch experiments on the denitrifying degradation of dimethyl phthalate (DMP) was most favorable at pH 7-9 and 30-35 degrees C. DMP was first degraded to monomethyl phthalate (MMP), which was in turn degraded to phthalate before complete mineralization.