Skip to Content
Merck
All Photos(2)

Key Documents

36732

Supelco

1-Naphthylacetamide

PESTANAL®, analytical standard

Synonym(s):

1-Naphthaleneacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CONH2
CAS Number:
Molecular Weight:
185.22
Beilstein:
1945547
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

180-183 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=O)Cc1cccc2ccccc12

InChI

1S/C12H11NO/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H2,13,14)

InChI key

XFNJVKMNNVCYEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ma Claudia Castañeda-Saucedo et al.
Plants (Basel, Switzerland), 9(3) (2020-03-04)
Stevia is an important non-caloric sweetener that has health-beneficial properties. The objective is to evaluate growth, development, and rooting of stevia plants during different seasons of the year using growth hormones. Eight experiments were set up in Ciudad Guzman, Jalisco
Yoshitaka Arai et al.
Journal of plant research, 117(3), 191-198 (2004-03-26)
The activity of indole-3-acetamide (IAM) hydrolase from rice cells was enriched ca. 628-fold by gel filtration and anion exchange column chromatography. The molecular masses of the IAM hydrolase estimated by gel filtration and sodium dodecyl sulfate polyacrylamide gel electrophoresis were
J Shah et al.
Molecular plant-microbe interactions : MPMI, 10(1), 69-78 (1997-01-01)
Salicylic acid (SA) plays an important signaling role in the resistance of many plants to pathogen invasion. Increases in endogenous SA levels have been associated with the hypersensitive response as well as systemic acquired resistance (SAR). SA also induces the
W P Cochrane et al.
Journal - Association of Official Analytical Chemists, 63(1), 145-148 (1980-01-01)
A simple, rapid, and accurate method has been developed for the determination of naphthalene-acetamide (NAAmide) residues in apples. After extraction with chloroform, separation is performed on an RP-8 high pressure liquid chromatographic column using acetonitrile-water (30 + 70) buffered to
C Ravikumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(2), 859-866 (2010-01-06)
FT Raman and IR spectra of the biologically active molecule, 1-naphthalene acetamide (NA) have been recorded and analyzed. The equilibrium geometry, bonding features and harmonic vibrational wavenumbers of NA have been calculated with the help of B3LYP density functional theory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service