Skip to Content
Merck
All Photos(1)

Key Documents

36192

Supelco

Phenmedipham

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O4
CAS Number:
Molecular Weight:
300.31
Beilstein:
2395027
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)Nc1cccc(OC(=O)Nc2cccc(C)c2)c1

InChI

1S/C16H16N2O4/c1-11-5-3-6-12(9-11)18-16(20)22-14-8-4-7-13(10-14)17-15(19)21-2/h3-10H,1-2H3,(H,17,19)(H,18,20)

InChI key

IDOWTHOLJBTAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenmedipham is a carbanilate herbicide, used primarily to control broad-leaved weeds in sugar beet harvest and spinach.

Application

Phenmedipham may be used as a reference standard for the determination of the analyte in oranges by high-performance liquid chromatography-triple-quadrupole mass spectrometry in combination with QuEChERS method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Voltammetric determination of phenmedipham herbicide using a multiwalled carbon nanotube paste electrode.
Demir E and Inam R
Turkish Journal of Chemistry, 42(4), 997-1007 (2018)
Sara C Novais et al.
Chemosphere, 79(2), 233-237 (2010-02-06)
Enchytraeids are important inhabitants of a wide variety of soil types and suitable test organisms for environmental risk assessment through the determination of effects on survival and reproduction. In this study, the effect of pesticides with different modes of action
Sara C Novais et al.
Ecotoxicology (London, England), 21(1), 213-224 (2011-09-06)
The soil oligochaete Enchytraeus albidus is a standard test organism used in biological testing for Environmental Risk Assessment (ERA). Although effects are known at acute and chronic level through survival, reproduction and avoidance behaviour endpoints, very little is known at
Thomas Müller et al.
Pest management science, 58(12), 1243-1249 (2002-12-13)
Ethoxylates of rapeseed oil and of methylated rapeseed oil were synthesized and tested as adjuvants for 2,4-D and phenmedipham. Provided they had less than 6 units of ethylene oxide (EO), 1.0 to 10 g litre(-1) ethoxylates in water induced droplet
Mónica J B Amorim et al.
Chemosphere, 61(8), 1102-1114 (2005-11-03)
Soil ecotoxicology studies are usually performed in standard soils, such as the OECD (Organization for Economic Cooperation and Development) artificial soil or the LUFA St. 2.2, a natural soil. When assessing the toxic effects in the environment, soil properties are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service