Skip to Content
Merck
All Photos(1)

Key Documents

35836

Supelco

4-Nitrophenol

PESTANAL®, analytical standard

Synonym(s):

p-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein:
1281877
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39032018
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.6 mmHg ( 120 °C)

product line

PESTANAL®

Assay

100% (HPLC)

autoignition temp.

541 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

279 °C (lit.)

mp

110-115 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

336.2 °F - closed cup

Flash Point(C)

169.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuan-I Chen et al.
Journal of agricultural and food chemistry, 60(51), 12540-12546 (2012-11-30)
A catalytic system for deglycosylation of isoflavone in black soybean milk was established. β-Glucosidase which was covalently immobilized onto the glass microspheres exhibited a significant efficiency for the conversion of pNPG to p-nitrophenol over other carriers. The optimum temperature for
Siew Hong Lam et al.
Biochimica et biophysica acta, 1830(10), 4778-4789 (2013-06-25)
4-Nitrophenol (4-NP) is a prioritized environmental pollutant and its toxicity has been investigated using zebrafish, advocated as an alternative toxicological model. However, molecular information of 4-NP induced hepatotoxicity is still limited. This study aimed to obtain molecular insights into 4-NP-induced
Simon De Corte et al.
Colloids and surfaces. B, Biointerfaces, 102, 898-904 (2012-10-31)
An increasing demand for catalytic Pd nanoparticles has motivated the search for sustainable production methods. An innovative approach uses bacterial cells as support material for synthesizing Pd nanoparticles by reduction of Pd(II) with e.g. hydrogen or formate. Nevertheless, drawbacks of
Jau-Rung Chiou et al.
Journal of hazardous materials, 248-249, 394-400 (2013-02-19)
Silver/iron oxide composite nanoparticles have been synthesized successfully via a facile one-pot green route by the use of l-arginine, which created an aqueous solution of about pH 10 and acted as a reducing agent for the successive formation of iron
Ipsita K Sen et al.
Carbohydrate polymers, 91(2), 518-528 (2012-11-06)
Gold nanoparticles were synthesized by reducing chloroauric acid with a glucan, isolated from an edible mushroom Pleurotus florida, cultivar Assam Florida. Here, glucan acts as reducing as well as stabilizing agent. The synthesized gold nanoparticles were characterized by UV-visible spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service