Skip to Content
Merck
All Photos(1)

Key Documents

35660

Sigma-Aldrich

Bis(2-chloroethyl) ether

puriss., ≥99.0% (GC)

Synonym(s):

2,2′-Dichlorodiethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2)2O
CAS Number:
Molecular Weight:
143.01
Beilstein:
605317
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.4 mmHg ( 20 °C)

Quality Level

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.456 (lit.)
n20/D 1.457

bp

65-67 °C/15 mmHg (lit.)

mp

−47 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

functional group

chloro
ether

SMILES string

ClCCOCCCl

InChI

1S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2

InChI key

ZNSMNVMLTJELDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-chloroethyl) ether is a chemical intermediate. 2-Chloroethyl β-D-glucosiduronic acid is the major metabolite of bis(2-chloroethyl)ether.

Application

Bis(2-chloroethyl) ether may be used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thiodiglycolic acid: a major metabolite of bis(2-chloroethyl)ether.
R D Lingg et al.
Toxicology and applied pharmacology, 47(1), 23-34 (1979-01-01)
Macrocyclic Polyethers: Dibenzo-18-crown-6 Polyether and Dicyclohexyl-18-crown-6 Polyether.
Pedersen CJ.
Organic Syntheses, 66-66 (1972)
Maria D M Mutuc et al.
Chemosphere, 70(8), 1390-1398 (2007-12-07)
This study examined the feasibility of using surface catalyzed Fenton treatment to remediate soil and groundwater contaminated by the chlorinated ethers, bis(2-chloroethyl) ether (BCEE) and bis(2-chloroethoxy) methane (BCEM). Parameters that affect the contaminant loss rate such as porewater pH, hydrogen
K Norpoth et al.
Journal of cancer research and clinical oncology, 112(2), 125-130 (1986-01-01)
Either 40 mumole or 160 mumole 2,2'-DDE was injected into male Wistar rats and the metabolites, TdGA and HEMA, were determined in the 24-h urine specimens. Comparative investigations were carried out giving equimolar amounts of chloroethanol and 2-chloroacetaldehyde diethyl acetal.
Kevin McClay et al.
Applied and environmental microbiology, 73(21), 6870-6875 (2007-09-18)
Degradation of bis(2-chloroethyl) ether (BCEE) was observed to occur in two bacterial strains. Strain ENV481, a Xanthobacter sp. strain, was isolated by enrichment culturing of samples from a Superfund site located in the northeastern United States. The strain was able

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service