Skip to Content
Merck
All Photos(2)

Documents

35601

Supelco

PCB No 28

analytical standard

Synonym(s):

2,4,4′-Trichlorobiphenyl, 2,4,4′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54
Beilstein:
1956846
Ballschmiter Number:
28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(cc1)-c2ccc(Cl)cc2Cl

InChI

1S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-6-5-10(14)7-12(11)15/h1-7H

InChI key

BZTYNSQSZHARAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls (PCBs) are groups of chemical compounds which are persistent in the environment and are considered highly toxic to humans and animals. PCB No 28 may be used for environmental analysis and precise quality control of food and feed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Li Xu et al.
Huan jing ke xue= Huanjing kexue, 31(1), 255-259 (2010-03-25)
The capability of Rhizobium meliloti on degrading 2,4,4'-trichlorobiphenyl and 18 kinds of polychlorinated biphenyl congener mixtures was studied by shaking flask experiment. The results showed that the degradation capability of Rhizobium meliloti to 2, 4, 4'-trichlorobiphenyl increased gradually with the
Xiang-Hui Sun et al.
Huan jing ke xue= Huanjing kexue, 31(10), 2327-2330 (2011-01-15)
Sorption behaviors of PCB28 to phosphatidylcholine (PC)and triglyceride (TG) were studied. Results showed that sorption equilibrium could be achieved in 8 h for PCB28, and the sorption amount on PC was higher than that of TG when the initial PCB28
Stuart Harrad et al.
Journal of environmental monitoring : JEM, 9(4), 314-318 (2007-04-06)
The environmentally prevalent polybrominated diphenyl ether (PBDE) #47 and polychlorinated biphenyls (PCBs) #28 and #118 were challenged for 24 hours with a novel biomass-supported Pd catalyst (Bio-Pd(0)). Analysis of the products via GC-MS revealed the Bio-Pd(0) to cause the challenged
Yu Wang et al.
Environmental science and pollution research international, 19(2), 448-457 (2011-08-09)
Dechlorination of polychlorinated biphenyls (PCBs) by nanoscale zerovalent iron (NZVI) is often strongly hindered by increased pH because large amounts of H(+) ions were consumed during the surface reaction. The main objective of this work was to evaluate the effect
Shui-Ping Wu et al.
Huan jing ke xue= Huanjing kexue, 32(11), 3277-3283 (2012-02-03)
The levels and congener patterns of 28 PCBs compounds were investigated in soil and dust fallout collected in a capacitor storage site and an industrial brownfield, respectively in Sichuan Ziyang Locomotive Factory. The highest concentration of the total PCBs(sigma PCBs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service