Skip to Content
Merck
All Photos(1)

Documents

35410

Supelco

Citrinin solution

~100 μg/mL in acetonitrile, analytical standard

Synonym(s):

4,6-Dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3H-2-benzopyran-7-carboxylic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O5
CAS Number:
Molecular Weight:
250.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1OC=C2C(O)=C(C(O)=O)C(=O)C(C)=C2[C@@H]1C

InChI

1S/C13H14O5/c1-5-7(3)18-4-8-9(5)6(2)11(14)10(12(8)15)13(16)17/h4-5,7,15H,1-3H3,(H,16,17)/t5-,7-/m1/s1

InChI key

CQIUKKVOEOPUDV-IYSWYEEDSA-N

General description

Citrinin is a hepato-nephrotoxic mycotoxin. It can be produced by several filamentous fungi genera of Monauscus, Aspergillus, and Penicillium, which is commonly found as a contaminant in stored grains, foods, feedstuffs and also in biological fluids.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual synergistic actions of silver nanoparticles with natural products on Ochratoxin A production
TM GA, et al.
Journal of Life Sciences, 14(3) (2017)
The Preparation and Identification of a Monoclonal Antibody against Citrinin and the Development of Detection via Indirect Competitive ELISA
Yirga KS, et al.
Toxins, 9(3), 110-110 (2017)
Presence of citrinin in grains and its possible health effects
Culig B, et al.
African Journal of Traditional, Complementary, and Alternative Medicines : AJTCAM / African Networks on Ethnomedicines, 14(3), 22-22 (2017)
Review on the qualitative and quantitative analysis of the mycotoxin citrinin
Xu J-B, et al.
Food Control, 17(4), 271- 285 (2006)
Hediyeh Davoudi Moghadam et al.
Journal of the science of food and agriculture, 99(9), 4439-4444 (2019-03-14)
Today, there is an increasing concern about the consumption of synthetic colorants in food because of their possible health hazards. Monascus purpureus has attracted a great deal of attention as it produces various coloured pigments with high chemical stability, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service