Skip to Content
Merck
All Photos(2)

Key Documents

34311

Supelco

Acifluorfen

PESTANAL®, analytical standard

Synonym(s):

5-(2-Chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoic acid, 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7ClF3NO5
CAS Number:
Molecular Weight:
361.66
Beilstein:
2953865
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C14H7ClF3NO5/c15-10-5-7(14(16,17)18)1-4-12(10)24-8-2-3-11(19(22)23)9(6-8)13(20)21/h1-6H,(H,20,21)

InChI key

NUFNQYOELLVIPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akifumi Sugiyama et al.
Plant & cell physiology, 46(8), 1428-1432 (2005-06-07)
Homoglutathione (hGSH), which is present in some leguminous plants, is preferred over GSH in in vitro conjugation of acifluorfen and fomesafen by glutathione S-transferase. To investigate the function of hGSH in in vivo detoxification of xenobiotics, we evaluated herbicide tolerance
Olga Soldatova et al.
Molecular genetics and genomics : MGG, 273(4), 311-318 (2005-04-09)
Several Arabidopsis mutants of the ecotype Dijon were isolated that show resistance to the herbicide acifluorfen, which inactivates protoporphyrinogen oxidase (PPOX), an enzyme of tetrapyrrole biosynthesis. This enzyme provides protoporphyrin for both Mg chelatase and ferrochelatase at the branchpoint, which
Alexey A Apchelimov et al.
Planta, 225(4), 935-943 (2006-11-24)
One of the key regulatory enzymes of the chlorophyll biosynthesis pathway is magnesium (Mg) chelatase, consisting of three different subunits CHLI, CHLD and CHLH. While CHLH and CHLD are encoded by a single gene each in Arabidopsis, CHLI is encoded
Koenraad F M J Tierens et al.
The Plant journal : for cell and molecular biology, 29(2), 131-140 (2002-02-28)
An Arabidopsis thaliana mutant, esa1, that shows enhanced susceptibility to the necrotrophic pathogens Alternaria brassicicola, Botrytis cinerea and Plectosphaerella cucumerina, but has wild-type levels of resistance to the biotrophic pathogens Pseudomonas syringae pv. tomato and Peronospora parasitica. The enhanced susceptibility
Lu Sun et al.
Amino acids, 37(3), 523-530 (2009-03-07)
Protoporphyrinogen IX oxidase (PPO), the last common enzyme of heme and chlorophyll biosynthesis, catalyses the oxidation of protoporphyrinogen IX to protoporphyrin IX, with FAD as cofactor. Among PPO, Bacillus subtilis PPO (bsPPO) is unique because of its broad substrate specificity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service