Skip to Content
Merck
All Photos(1)

Key Documents

33420

Sigma-Aldrich

1-Naphthol

puriss. p.a., reag. Ph. Eur., ≥99% (GC)

Synonym(s):

α-Naphthol, 1-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
1817321
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

vapor density

4.5 (120 °C, vs air)

vapor pressure

1 mmHg ( 94 °C)

grade

puriss. p.a.

Assay

≥99% (GC)

autoignition temp.

1007 °F

expl. lim.

5 %

impurities

≤0.001% heavy metals (as Pb)
≤0.2% naphthaline (GC)
≤0.2% water (Karl Fischer)
≤0.5% 2-naphthol (GC)

ign. residue

≤0.05% (as SO4)

bp

278-280 °C (lit.)

mp

94-96 °C (lit.)
94-97 °C

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

Oc1cccc2ccccc12

InChI

1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H

InChI key

KJCVRFUGPWSIIH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application


  • A ′conovenomic′ analysis of the milked venom from the mollusk-hunting cone snail Conus textile--the pharmacological importance of post-translational modifications.: This research underscores the critical role of post-translational modifications in the biological activity of peptides derived from cone snail venom, illustrating the potential for developing novel pharmacological agents using peptide synthesis techniques like those involving Fmoc-Asp(OtBu)-OH (Bergeron et al., 2013).

  • Parallel high-throughput accurate mass measurement using a nine-channel multiplexed electrospray liquid chromatography ultraviolet time-of-flight mass spectrometry system.: This paper discusses a breakthrough in mass spectrometry, highlighting a method for high-throughput analysis which could be critical for the characterization of complex peptide products synthesized using Fmoc-Asp(OtBu)-OH (Fang et al., 2003).


Packaging

bottled under inert gas

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accurate hydrogen-bonding energies between 1-naphthol and water, methanol and ammonia.
Burgi T, et al
Chemical Physics Letters, 246(3), 291-299 (1995)
Baoliang Chen et al.
Chemosphere, 76(1), 127-133 (2009-03-14)
Biochars, derived from biomass, are increasingly recognized as an environmental-friendly sorbent to abate organic pollutants. Sorption variations of biochars with their pyrolytic temperatures are evaluated. Nine biochars of orange peels with different pyrolytic temperatures (150-700 degrees C, referred as OP150-OP700)
Characterization of duloxetine HCL API and its process related impurities
Karagiannidou E, et al.
International Journal of Analytical, Pharmaceutical and Biomedical Sciences, 3(2) (2014)
Anika Kasprick et al.
European journal of immunology, 45(5), 1462-1470 (2015-02-14)
The role of mast cells (MCs) in autoimmunity is the matter of an intensive scientific debate. Based on observations in different MC-deficient mouse strains, MCs are considered as fundamental players in autoimmune diseases. However, most recent data suggest that the
Shitong Yang et al.
The Science of the total environment, 420, 214-221 (2012-02-15)
The increasing exploitation of multi-walled carbon nanotubes (MWCNTs) into many industrial processes has raised considerable concerns as they are likely to be released into the environment. The interactions of the pollutants in water with discharged MWNCTs will further influence the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service