Skip to Content
Merck
All Photos(1)

Key Documents

33372

Supelco

Nitrapyrin

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-6-(trichloromethyl)pyridine, CP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl4N
CAS Number:
Molecular Weight:
230.91
Beilstein:
1618997
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

61-65 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cccc(n1)C(Cl)(Cl)Cl

InChI

1S/C6H3Cl4N/c7-5-3-1-2-4(11-5)6(8,9)10/h1-3H

InChI key

DCUJJWWUNKIJPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrapyrin is a nitrification inhibitor, which can also undergo hydrolysis to form 6-chloropicolinic acid, that can inhibit methane oxidation by M. trichosporium OB3b.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of nitrapyrin and chloropicolinic acid on ammonium oxidation by Nitrosomonas europaea
Powell, S.J.; JProsser, J.I.;
FEMS Microbiology Letters, 28, 51-54 (1985)
G W Randall et al.
Journal of environmental quality, 32(5), 1764-1772 (2003-10-11)
Subsurface drainage, a water management practice used to remove excess water from poorly drained soils, can transport substantial amounts of NO3 from agricultural crop production systems to surface waters. A field study was conducted from the fall of 1986 through
E Kostyál et al.
Applied microbiology and biotechnology, 50(5), 612-622 (1998-12-29)
The dechlorinating and genotoxicity-removing activities of nitrifying fluidized-bed reactor biomass towards chlorinated organic compounds in water were shown at level below 1 ppm. The removal rates of adsorbable organic halogens were 200 micrograms Cl (g VS day)-1 for chlorinated humic
D Pagliaccia et al.
Microbial ecology, 56(3), 538-554 (2008-03-19)
Fluorescent pseudomonads have been associated, via diverse mechanisms, with suppression of root disease caused by numerous fungal and fungal-like pathogens. However, inconsistent performance in disease abatement, after their employment, has been a problem. This has been attributed, in part, to
Effect of glyphosate and nitrapyrin on selected bacterial populations in continuous-flow culture.
C W Hendricks et al.
Bulletin of environmental contamination and toxicology, 49(3), 417-424 (1992-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service