Skip to Content
Merck
All Photos(1)

Key Documents

33254

Supelco

4-tert-Octylphenol-3,5-d2 diethoxylate solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxyl-3,5-d2]ethoxy}ethanol, 4-tert-OP2EO-D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D2H28O3
CAS Number:
Molecular Weight:
296.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

[2H]c1cc(OCCOCCO)cc([2H])c1C(C)(C)CC(C)(C)C

InChI

1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3/i6D,7D

InChI key

LBCZOTMMGHGTPH-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

4-tert-Octylphenol-3,5-d2 diethoxylate is an analytical standard used for the quantification of mono and di-ethoxylates of nonylphenol (NP) and octylphenol (OP) in environmental samples using solid-phase micro extraction followed by gas chromatography with mass spectrometry (SPME-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service