Skip to Content
Merck
All Photos(1)

Key Documents

323764

Sigma-Aldrich

1,1′-Diethyl-2,2′-cyanine iodide

97%

Synonym(s):

1-Ethyl-2-[(1-ethyl-2(1H)-quinolinylidene)methyl]quinolinium iodide, Decynium 22, Pseudocyanine iodide, Pseudoisocyanine iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23IN2
CAS Number:
Molecular Weight:
454.35
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97%

form

powder or crystals

mp

273 °C (dec.) (lit.)

λmax

524 nm

ε (extinction coefficient)

≥25000 at 487-495 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[I-].CCN1\C(C=Cc2ccccc12)=C\c3ccc4ccccc4[n+]3CC

InChI

1S/C23H23N2.HI/c1-3-24-20(15-13-18-9-5-7-11-22(18)24)17-21-16-14-19-10-6-8-12-23(19)25(21)4-2;/h5-17H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

GMYRVMSXMHEDTL-UHFFFAOYSA-M

General description

1,1′-Diethyl-2,2′-cyanine iodide is a quinocyanine dye. This photographic sensitizing dye is mainly used in silver halide photography. 1,1′-Diethyl-2,2′-cyanine iodide dye aggregation in dilute aqueous solution displays red and blue shifted absorption bands.

Application

1,1′-Diethyl-2,2′-cyanine iodide has been used as an absorber in a method to correct the fluorescence excitation-emission matrix (EEM).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yuichi Hashimoto et al.
Journal of colloid and interface science, 311(1), 289-295 (2007-03-21)
We demonstrate a simple method for fabricating a two-dimensional array of microdomes that consist of cyanine dye complexes. Investigation of the morphology and the fluorescence emission of microdomes was carried out before/after annealing. The principal of microdome formation is "dewetting,"
Ah-Young Jee et al.
Physical chemistry chemical physics : PCCP, 13(33), 15227-15232 (2011-07-20)
The trans-cis isomerization of an excited molecule converts light energy into mechanical motion, which interacts cooperatively with its surroundings. To understand such a photodynamic process in solids, we investigated the internal twisting motion of 1,1'-diethyl-2,2'-cyanine iodide (DCI) in a series
Gary C H Mo et al.
Langmuir : the ACS journal of surfaces and colloids, 25(18), 10719-10729 (2009-08-04)
Controlling the self-assembly of molecules into specific structural motifs has important implications for the design of materials with specific optical properties. We report here the results of a correlated confocal fluorescence-atomic force microscopy (AFM) study of pseudoisocyanine iodide (PIC) self-assembly
Jonathan E Hill et al.
Physiology & behavior, 104(2), 306-311 (2010-11-18)
In addition to exerting actions via mineralocorticoid and glucocorticoid receptors, corticosteroids also act by inhibiting uptake(2), a high-capacity monoamine transport system originally described in peripheral tissues. Recent studies have demonstrated that uptake(2) transporters are expressed in the brain and play
Mustafa M Demir et al.
The journal of physical chemistry. B, 113(34), 11568-11573 (2009-10-23)
Submicrometer diameter, light emitting fibers of poly(vinyl alcohol) (PVA) doped with pseudoisocyanine (1,1'-diethyl-2,2'-cyanine bromide, PIC) dye were prepared by electrospinning. A horizontal setup was employed with a stationary collector consisting of two parallel-positioned metal strips separated by a void gap.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service