Skip to Content
Merck
All Photos(1)

Key Documents

32301-M

Sigma-Aldrich

Ammonium acetate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
solid

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

<0.001 hPa

Assay

≥98%

form

solid

impurities

≤0.0002% heavy metals (as Pb)
≤0.005% KMnO4 red. matter (as HCOOH)
≤2% water (Karl Fischer)

ign. residue

≤0.01% (as SO4)

pH

6.5-7.5 (25 °C, 77.1 g/L)

mp

110-112 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤10 ppm
Cd: ≤2 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤2 ppm
Fe: ≤2 ppm
K: ≤50 ppm
Mg: ≤2 ppm
Mn: ≤5 ppm
Na: ≤50 ppm
Ni: ≤5 ppm
Pb: ≤2 ppm
Zn: ≤2 ppm

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium acetate is a colorless hygroscopic solid. It can be prepared by reacting glacial acetic acid with ammonia or ammonium carbonate. It has been reported as potential inhibitor of Pd/C mediated hydrogenolysis of benzyl ether.

Application

Ammonium acetate has been used in the mobile phase for the HPLC determination of ezetimibe and its metabolites. It may be used in preparation of phenanthrimidazoles and retinimidazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective inhibition of benzyl ether hydrogenolysis with Pd/C due to the presence of ammonia, pyridine or ammonium acetate.
Sajiki H.
Tetrahedron Letters, 36(20), 3465-3468 (1995)
Reactions of phenanthraquinone and retenequinone with aldehydes and ammonium acetate in acetic acid solution1.
Steck EA and Day AR.
Journal of the American Chemical Society, 65(3), 452-456 (1943)
Anima Ghosal et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(3), 314-320 (2004-02-24)
Ezetimibe [1-(4-fluorophenyl)-3(R)-[3-(4-fluorophenyl)-3(S)-hydroxypropyl]-4(S)-(4-hydroxyphenyl)-2-azetidinone] (Zetia; Schering-Plough, Kenilworth, NJ) is the first in a new class of cholesterol-lowering agents known as cholesterol absorption inhibitors. The objective of this study was to identify the isoform(s) of human liver and intestinal UDP-glucuronosyltransferase (UGT) enzymes responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service