Skip to Content
Merck
All Photos(1)

Documents

32012

Supelco

Metalaxyl

PESTANAL®, analytical standard

Synonym(s):

N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-DL-alanine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
Beilstein:
2947777
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

69-73 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

COCC(=O)N(C(C)C(=O)OC)c1c(C)cccc1C

InChI

1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3

InChI key

ZQEIXNIJLIKNTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metalaxyl, also known as Ridomil, is a phenylamide and systemic benzoic fungicide. It is generally used to control infection caused by Phytophthora infestans.

Application

Metalaxyl has been used as reference standard in Fourier transform infrared (FTIR) spectrometric method for determination of metalaxyl in pesticide formulations.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous determination of Folpet and Metalaxyl in pesticide formulations by flow injection Fourier transform infrared spectrometry.
Quintas, Guillermo, et al.
Analytica Chimica Acta, 480.1, 11-21 (2003)
Widespread distribution and probable origin of resistance to metalaxyl in clonal genotypes of Phytophthora infestans in the United States and Western Canada.
Goodwin, Stephen B., Ludwik S. Sujkowski, and William E. Fry.
Phytopathology, 86, 793-800 (1996)
R Celis et al.
The Science of the total environment, 444, 288-297 (2013-01-02)
Improving the existing knowledge on the enantioselectivity of processes affecting chiral pesticide enantiomers in the environment is necessary to maximize the efficacy and minimize the environmental impact caused by the use of pesticides with chiral properties. In this work, the
R Sowmya et al.
Marine biotechnology (New York, N.Y.), 13(5), 918-927 (2011-01-19)
Studies were carried out to utilize in situ proteases of shrimp heads to recover carotenoproteins possessing antioxidant activity. Highest protease activity of the buffer extract was found at pH 8.0 (9.85 ± 0.61 units). The protease activity increased with temperature up to
Miao Han et al.
The Journal of eukaryotic microbiology, 60(1), 79-88 (2012-12-01)
The genetic diversity of 85 isolates of Phytophthora infestans collected in 2007 from Gansu province in China was determined and compared with 21 isolates collected before 2004. Among them, 70 belonged to the A1 mating type and 15 were self-fertile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service