Skip to Content
Merck
All Photos(1)

Documents

30885

Supelco

Thiourea

≥99.999% (metals basis)

Synonym(s):

Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥99.999% (metals basis)

mp

170-176 °C (lit.)

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Au: ≤0.01 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.05 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤2 mg/kg
Cd: ≤0.01 mg/kg
Ce: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.05 mg/kg
Cs: ≤0.01 mg/kg
Cu: ≤0.01 mg/kg
Dy: ≤0.01 mg/kg
Er: ≤0.01 mg/kg
Eu: ≤0.01 mg/kg
Fe: ≤0.5 mg/kg
Ga: ≤0.01 mg/kg
Gd: ≤0.01 mg/kg
Ge: ≤0.01 mg/kg
Hf: ≤0.01 mg/kg
Hg: ≤0.005 mg/kg
Ho: ≤0.01 mg/kg
In: ≤0.01 mg/kg
Ir: ≤0.01 mg/kg
K: ≤0.2 mg/kg
La: ≤0.01 mg/kg
Li: ≤0.01 mg/kg
Lu: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.01 mg/kg
Na: ≤0.5 mg/kg
Nb: ≤0.01 mg/kg
Nd: ≤0.01 mg/kg
Ni: ≤0.05 mg/kg
Os: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pd: ≤0.05 mg/kg
Pr: ≤0.01 mg/kg
Pt: ≤0.05 mg/kg
Rb: ≤0.01 mg/kg
Re: ≤0.01 mg/kg
Rh: ≤0.01 mg/kg
Ru: ≤0.01 mg/kg
Sb: ≤0.01 mg/kg
Sc: ≤0.05 mg/kg
Se: ≤0.05 mg/kg
Sm: ≤0.01 mg/kg
Sn: ≤0.01 mg/kg
Sr: ≤0.1 mg/kg
Ta: ≤0.01 mg/kg
Tb: ≤0.01 mg/kg
Te: ≤0.01 mg/kg
Th: ≤0.01 mg/kg
Ti: ≤0.05 mg/kg
Tl: ≤0.01 mg/kg
Tm: ≤0.01 mg/kg
U: ≤0.01 mg/kg
V: ≤0.01 mg/kg
W: ≤0.01 mg/kg
Y: ≤0.01 mg/kg
Yb: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.05 mg/kg

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
Thiourea is a chaotropic agent; strong denaturant. Increases solubility and recovery of proteins.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of thiourea to increase the solubility of membrane proteins in two-dimensional electrophoresis.
Rabilloud T.
Electrophoresis, 19(5), 758-760 (1998)
Proteomic analysis of the human pathogen Trypanosoma cruzi.
Paba J, et al.
Proteomics, 4(4), 1052-1059 (2004)
Denaturation of Globular Proteins I. THE INTERACTION OF UREA AND THIOUREA WITH BOVINE PLASMA ALBUMIN.
Gordon JA and Warren JR
The Journal of Biological Chemistry, 243(21), 5663-5669 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service