26206
Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock
1725TLL, volume 250 μL, PTFE Luer lock (with slots), needle size (not included)
All Photos(2)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Recommended Products
material
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81127
needle L
, n/a
volume
250 μL
fitting
luer lock (without slots)
Related Categories
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been equipped in precision syringe pump of isothermal titration calorimeter which has been used in measuring the molar enthalpies of binary mixtures containing 2-decanone or dipentyl ether. It has also been used in the synthesis of single-molecule DNA by DNA polymerase with microflow channels.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Pharmacology for the Prehospital Professional, 54-54 (2009)
A New Direct Single-Molecule Observation Method for DNA Synthesis Reaction Using Fluorescent Replication Protein A.
Takahashi, Shunsuke, et al.
Sensors, 14.3, 5174-5182 (2014)
Excess molar enthalpies of binary mixtures containing 2-decanone or dipentyl ether with long-chain n-alkanes at T= 298.15 K.
Liao, Wei-Chen, Ho-mu Lin, and Ming-Jer Lee
The Journal of Chemical Thermodynamics, 43.4, 515-520 (2011)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)