This plunger is 304 stainless steel coated with PTFE. Yes, there is a PTFE plunger tip on this syringe to help create the gas-tight seal.
20889
Hamilton® HPLC syringes
1725RNR, volume 250 μL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.)
About This Item
Recommended Products
material
blunt tip stainless steel needle (point style 3)
glass body
needle size
22 ga (blunt tip)
feature
blunt tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81165
needle L
51 mm (2 in.)
volume
250 μL
compatibility
for use with Rheodyne
for use with SSI
for use with Valco VISF-2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
Which material is the plunger made of? Does it have a rubber end like many Hamilton syringes?
1 answer-
Helpful?
-