Skip to Content
Merck
All Photos(1)

Key Documents

09651

Sigma-Aldrich

1-Chlorobutane

biotech. grade, for protein sequence analysis, ≥99.8%

Synonym(s):

Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Cl
CAS Number:
Molecular Weight:
92.57
Beilstein:
1730909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

biotech. grade
for protein sequence analysis

Quality Level

vapor density

3.2 (vs air)

vapor pressure

80.1 mmHg ( 78.4 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

860 °F

expl. lim.

10.1 %

impurities

≤0.01% water (Karl Fischer)

evapn. residue

≤0.0003%

refractive index

n20/D 1.402 (lit.)
n20/D 1.402

bp

77-78 °C (lit.)

mp

−123 °C (lit.)

density

0.886 g/mL at 25 °C (lit.)

suitability

corresponds for Infrared spectrum
in accordance for sequence analysis test

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CCCCCl

InChI

1S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3

InChI key

VFWCMGCRMGJXDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Special packaging for direct use in common sequencers; contains 200 ml.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hazel M Rivera et al.
European journal of mass spectrometry (Chichester, England), 9(6), 599-607 (2004-04-22)
A sensitive and selective high-performance liquid chromatography/atmospheric pressure chemical ionisation tandem mass spectrometry (HPLC/APCI-MS/MS) method for the simultaneous detection of 18 benzodiazepines and metabolites in human blood is described. The procedure utilises butyl chloride extraction at alkaline pH followed by
Javier Catalán et al.
Physical chemistry chemical physics : PCCP, 13(33), 15022-15030 (2011-07-16)
In this work, we substantiate the change in the emitting state of indole caused by the dipolarity increase in the solvent 1-chlorobutane, observed on lowering the temperature from 293 to 133 K, accompanied by no significant changes in the corresponding
Antonio García Martínez et al.
The Journal of organic chemistry, 70(25), 10238-10246 (2005-12-06)
[structure: see text] The activation energy in the gas phase (deltaE(double dagger)) and the free energy of activation (deltaG(double dagger)) in water solution for the hydrolysis of the monohydrates of methyl chloride (MeCl), tert-butyl chloride (t-BuCl), and 1-adamantyl chloride (AdCl)
Benjamin Erable et al.
Biotechnology and bioengineering, 86(1), 47-54 (2004-03-10)
Biofiltration of air polluted by volatile organic compounds is now recognized by the industrial and research communities as an effective and viable alternative to standard environmental technologies. Whereas many studies have focused on solid/liquid/gas biofilters, there have been fewer reports
D E Moody et al.
Journal of analytical toxicology, 19(6), 343-351 (1995-10-01)
l-alpha-Acetylmethadol (LAAM) is approved as a substitute for methadone for the treatment of opiate addiction. Analytical methods are needed to quantitate LAAM and its two psychoactive metabolites, norLAAM and dinorLAAM, to support pharmacokinetic and other studies. We developed a gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service