Skip to Content
Merck
All Photos(2)

Key Documents

08198

Supelco

Eleutheroside E

analytical standard

Synonym(s):

[(1R,3aR,4S,6aS)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2,6-dimethoxy-4,1-phenylene) bis-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H46O18
CAS Number:
Molecular Weight:
742.72
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3OC[C@@H]4[C@@H]3CO[C@H]4c5cc(OC)c(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c(OC)c5

InChI

1S/C34H46O18/c1-43-17-5-13(6-18(44-2)31(17)51-33-27(41)25(39)23(37)21(9-35)49-33)29-15-11-48-30(16(15)12-47-29)14-7-19(45-3)32(20(8-14)46-4)52-34-28(42)26(40)24(38)22(10-36)50-34/h5-8,15-16,21-30,33-42H,9-12H2,1-4H3/t15?,16?,21-,22+,23-,24+,25+,26-,27-,28+,29?,30?,33+,34-

InChI key

FFDULTAFAQRACT-RGFZIUCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eleutheroside E is one of the major bioactive saponins of Eleutherococcus senticosus.

Application

Eleutheroside E (ELU E) may be used as a reference standard for the analysis of ELU E in:
  • Rat plasma and tissue by solid-phase extraction (SPE) followed by high-performance liquid chromatography (HPLC) and photodiode array detection (PDA).
  • Acanthopanax senticosus by ionic liquids-ultrasound assisted extraction (ILUAE) followed by HPLC with ultraviolet (UV) detection.
  • Eleutherococcus senticosus Maxim. by rapid resolution liquid chromatography (RRLC) equipped with multi-wavelength UV detector.
  • Acanthopanax giraldii Harms by HPLC with diode array detector (DAD).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of Eleutheroside B and Eleutheroside E in rat plasma by high performance liquid chromatography-electrospray ionization mass spectrometry and its application in a pharmacokinetic study
Ma B, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 917, 84-92 (2013)
Semipreparative separation and determination of eleutheroside E in Acanthopanax giraldii Harms by high-performance liquid chromatography
Wang Z, et al.
Journal of Chromatographic Science, 43(5), 249-252 (2005)
L B Kardono et al.
Journal of natural products, 53(6), 1447-1455 (1990-11-01)
By bioactivity-directed fractionation, six cytotoxic constituents have been characterized from the bark of Plumeria rubra collected in Indonesia. Three iridoids, fulvoplumierin [1], allamcin [2], and allamandin [3], as well as 2,5-dimethoxy-p-benzoquinone [4], were found to be active constituents of the
D H Kim et al.
Archives of pharmacal research, 22(1), 30-34 (1999-03-11)
When liriodendrin or syringin was incubated for 24 h with human intestinal bacteria, two metabolites, (+)-syringaresinol-beta-D-glucopyranoside and (+)-syringaresinol, from liriodendrin and one metabolite, synapyl alcohol, from syringin were produced. The metabolic time course of liriodendrin was as follows: at early
Tomoko Sano et al.
Journal of natural medicines, 64(3), 257-265 (2010-03-11)
Boi and its original plant Sinomenium acutum from Japan were compared with Seifuto and its botanical origins from China in terms of their internal transcribed spacer (ITS) sequences and major chemical components. Boi, Seifuto, and their botanical origins overall showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service