Skip to Content

08062

Protein G′ from Streptococcus sp.

recombinant, expressed in E. coli

Synonym(s):

IgG F(c)-Receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
MDL number:
Clone:
-
Species reactivity:
-
Application:
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Streptococcus sp.)

Quality Segment

recombinant

expressed in E. coli

form

lyophilized solid

mol wt

~30 kDa

capacity

~5 mg/mg, solid binding capacity (IgG)

storage temp.

−20°C

General description

Genetically engineered truncated protein G; retains affinity for IgG, but lacks albumin and Fab binding sites and membrane-binding regions.
Protein G, a cell wall protein, is obtained from group G streptococci. The extracellular part of this protein is made of two/three small domains or serum albumin binding (GA domains) and two/three immunoglobulin (IgG) binding domains (B domains).

Application

Protein G′ from Streptococcus sp. has been used in in vitro actin labeling assay.

Biochem/physiol Actions

Protein G can bind all the human and mouse IgG subclasses. It can bind to both the fragment crystallizable (Fc) and antigen-binding fragment (Fab) components of the antibody.

Physical form

lyophilized powder in a Tris-HCl buffer, pH 7.5


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library