Skip to Content
Merck
All Photos(2)

Documents

07041

Supelco

Methyl laurate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Lauric acid methyl ester, Methyl dodecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CO2CH3
CAS Number:
Molecular Weight:
214.34
Beilstein:
1767780
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

200
300

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.431 (lit.)

bp

262 °C/766 mmHg (lit.)

mp

4-5 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)OC

InChI

1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3

InChI key

UQDUPQYQJKYHQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Komata et al.
Biological & pharmaceutical bulletin, 17(5), 705-708 (1994-05-01)
The effect of long chain fatty acid (FA) and its analogs on the accumulation of thiamine disulfide (TDS) in rat skin using propylene glycol as a vehicle was studied in vitro. Lauric acid (12:0) increased the accumulation of TDS in
D A Dimas et al.
AAPS PharmSciTech, 1(2), E16-E16 (2004-01-20)
The effects of coating thickness, type of adhesive, and type and concentration of enhancer on the mechanical properties of two acrylic pressure-sensitive adhesives (PSAs) were investigated using a 2(4) factorial design and an optimization technique. Sixteen formulations containing 0% or
E Barboza-Silva et al.
Oral microbiology and immunology, 24(4), 265-271 (2009-07-04)
The arginine deiminase system (ADS) of oral bacteria is a major generator of alkali (ammonia) in dental plaque and is considered to have anticaries effects. However, many of the antimicrobial agents used in oral care products may reduce alkali production
Jitender Sharma et al.
Journal of bioscience and bioengineering, 100(6), 662-666 (2006-02-14)
Efficient selective synthesis of the secondary amide surfactant N-methyl lauroylethanolamide from methyl laurate and N-methylethanol amine by carrier-fixed Chirazyme L-2 (Candida antarctica) using a kinetic strategy has been demonstrated. When different solvents were screened for product yields using Chirazyme L-2
Néstor M Carballeira et al.
Lipids, 40(10), 1063-1068 (2005-12-31)
The hitherto unknown 2-methylsulfanyldecanoic acid and 2-methylsulfanyldodecanoic acid were synthesized from methyl decanoate and methyl dodecanoate, respectively, through the reaction of lithium diisopropylamide and dimethyldisulfide in THF followed by saponification with potassium hydroxide in ethanol. Both alpha-methylsulfanylated FA were cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service