Skip to Content
Merck
All Photos(1)

Key Documents

03940

Sigma-Aldrich

2-Imidazolidinethione

purum, ≥98.0% (HPLC)

Synonym(s):

2-Thioxoimidazolidine, N,N′-Ethylenethiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
Beilstein:
106275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

biological source

synthetic

Quality Level

grade

purum

Assay

≥98.0% (HPLC)

form

powder

mp

196-200 °C (lit.)
196-200 °C

solubility

water: soluble 27.4 g/L at 20 °C

SMILES string

S=C1NCCN1

InChI

1S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

PDQAZBWRQCGBEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reiko Kaneko et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 53(1), 52-56 (2012-03-28)
An improved analysis method for 2-mercaptoimidazoline in rubber products containing chlorine was developed. 2-Mercaptoimidazoline (20 µg/mL) is detected by means of TLC with two developing solvents in the official method. But, this method is not quantitative. Instead, we employed HPLC
D Boers et al.
Human & experimental toxicology, 27(9), 721-727 (2008-12-02)
We conducted a multicenter prospective study to assess the effects of occupational exposure to ethylenebisdithiocarbamate fungicides and/or other pesticides on self-reported asthma and asthmatic symptoms. This multicenter study was conducted among 248 workers exposed to pesticides and 231 non-exposed workers
Huimin Jia et al.
The Journal of surgical research, 172(1), 131-136 (2010-10-05)
This study was designed to investigate the expression of Notch-1 and Jagged-2 in the terminal hindgut in ethylenethiourea (ETU)-exposed rat embryos with anorectal malformations (ARMs) and its potential association with the maldevelopment of the terminal hindgut in ARMs. ETU-exposed ARMs
Ethylene thiourea.
Report on carcinogens : carcinogen profiles, 12, 191-194 (2011-08-20)
Hongmei Pan et al.
Wei sheng yan jiu = Journal of hygiene research, 39(1), 79-82 (2010-04-07)
To investigate the effects of ethylenethiourea (ETU) on thyroglobulin (TG) secretion and iodide uptake of FRTL-5 cells. FRTL-5 cells were treated with 30, 150 and 270 microg/ml ETU, the cytotoxicity was tested by MTT and 3H-TdR, the synthesis and secretion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service