Skip to Content
Merck
All Photos(1)

Key Documents

03810585

Orientin

primary reference standard

Synonym(s):

8-β-D-Glucopyranosyl-3′,4′,5,7-tetrahydroxyflavone, Luteolin 8-C-β-D-glucopyranoside, Luteolin-8-glucoside, Lutexin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)c(O)c4

InChI

1S/C21H20O11/c22-6-14-17(28)18(29)19(30)21(32-14)16-11(26)4-10(25)15-12(27)5-13(31-20(15)16)7-1-2-8(23)9(24)3-7/h1-5,14,17-19,21-26,28-30H,6H2/t14-,17-,18+,19-,21+/m1/s1

InChI key

PLAPMLGJVGLZOV-VPRICQMDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Flavonoid glycoside found in the acai tree and globe and passion flowers. Antiviral, antimicrobial and antioxidant. Exhibits radioprotective activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Shagufta Perveen et al.
Journal of Asian natural products research, 13(9), 799-804 (2011-08-13)
Two new C-glycosylflavonoids celtisides A (1) and B (2) have been isolated from n-butanol-soluble fraction of Celtis africana, along with five known C-glycosylflavonoids vitexin (3), orientin (4), isoswertiajaponin (5), isoswertisin (6), and 2″-O-rhamnosyl vitexin (7) reported for the first time
P Uma Devi et al.
International journal of radiation biology, 80(9), 653-662 (2004-12-14)
To study the protective effect of orientin and vicenin against early genomic effects of foetal irradiation and their late consequences in mice. Fourteen-day pregnant mice were exposed to 1 Gy 60Co gamma-radiation 30 min after an intraperitoneal injection of orientin
Dalene de Beer et al.
Journal of the science of food and agriculture, 92(2), 274-282 (2011-07-23)
The effects of citric and ascorbic acids on the stability of aspalathin in rooibos (Aspalathus linearis) ready-to-drink (RTD) formulations containing fermented rooibos extract (FR), aspalathin-enriched green rooibos extract (GR) and aspalathin-enriched green rooibos extract ascorbic acid solubilisate (GR-solubilisate) were investigated
Nan Wu et al.
Molecules (Basel, Switzerland), 14(3), 1032-1043 (2009-03-24)
Antioxidant activities of the aqueous and ethanol extracts of pigeonpea [Cajanus cajan (L.) Millsp.] leaves, as well as petroleum ether, ethyl acetate, n-butanol and water fractions and the four main compounds separated from the ethanol extract, i.e. cajaninstilbene acid (3-hydroxy-4-prenylmethoxystilbene-2-carboxylic
Rosi Zanoni Da Silva et al.
Journal of natural medicines, 64(4), 402-408 (2010-05-18)
The antinociceptive properties of some fractions and two pure compounds, conocarpan and orientin, obtained from P. solmsianum leaves were investigated in several models of pain in mice. The results indicated that this plant exhibits a promising antinociceptive profile, as it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service