Skip to Content
Merck

LIBDH-RO

Roche

Liberase DH Research Grade

lyophilized, optimum pH 7.4, optimum reaction temp. 35-37 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.21
UNSPSC Code:
12352204

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Liberase DH Research Grade, lyophilized, optimum pH 7.4, optimum reaction temp. 35-37 °C

biological source

enzyme from Bacillus polymyxa (Protease)
enzyme from Clostridium histolyticum (collagenase)

description

high Dispase concentration

form

lyophilized

packaging

pkg of 10 mg (05401054001 [2 x 5 mg])
pkg of 100 mg (05401089001 [2 x 50 mg])

manufacturer/tradename

Roche

parameter

35-37 °C optimum reaction temp.

technique(s)

activity assay: suitable

color

white

optimum pH

7.4

solubility

water: soluble

application(s)

life science and biopharma

foreign activity

Endotoxin 3 EU/mg

shipped in

dry ice

storage temp.

−20°C

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
05401020001LIBTM-ROLIBTH-RO
biological source

enzyme from Bacillus polymyxa (Protease), enzyme from Clostridium histolyticum (collagenase)

biological source

Bacillus polymyxa (dispase), Bacillus sp. (thermolysin - Bacillus thermoproteolyticus), Clostridium histolyticum (collagenase)

biological source

-

biological source

Bacillus polymyxa (dispase), Bacillus sp. (thermolysin - Bacillus thermoproteolyticus), Clostridium histolyticum (collagenase)

technique(s)

activity assay: suitable

technique(s)

tissue processing: suitable

technique(s)

single cell analysis: suitable, tissue processing: suitable

technique(s)

tissue processing: suitable

application(s)

life science and biopharma

application(s)

-

application(s)

life science and biopharma
sample preparation

application(s)

-

form

lyophilized

form

lyophilized

form

lyophilized

form

lyophilized

shipped in

dry ice

shipped in

-

shipped in

dry ice

shipped in

-

optimum pH

7.4

optimum pH

7.4

optimum pH

7.4

optimum pH

7.4

Application

Liberase DH (Dispase High) Research Grade is used for the dissociation of a broad range of tissue types, where high purity of the enzyme blend is necessary for high cell yield and viability. Bacterial by-products, such as endotoxins, are reduced up to several thousand-fold.
It has been used for the digestion of metastatic tumor fragments.[1] It has been used for the enzymatic detachment of human embryonic stem cells colonies.[2]

Features and Benefits

  • Maximize viability and yield of isolated cells
with an enzyme blend that has less clostripain and trypsin activity, as well as reduced endotoxin content.
  • Count on higher specific activity of the enzyme blend
as a result of higher Collagenase I + II purity (determined by HPLC analysis).
  • Obtain higher experimental reproducibility
due to higher lot-to-lot consistency.

General description

The most commonly used method for decellularization procedures, such as tissue dissociation and cell harvesting, is based on the use of proteolytic enzymes. Proteases disrupt the extracellular matrix of tissue to allow the release of individual cells or the harvesting of cultured cells for transfer. The goal of cell isolation is to maximize the yield of dissociated cells that are viable and functionally active.[3]
Liberase enzyme technology comprises methods for purifying clostridial collagenase isoforms to high specific activity, and for blending them together with high-specific-activity neutral protease in optimal ratios for effective dissociation of primary tissues and cultured cells. Liberase DH Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a high concentration of Dispase®, a non-clostridial neutral protease.[3]
Liberase enzymes are designed to increase the quality and reproducibility of tissue dissociation, and improve the viability and functionality of isolated cells. Liberase Purified Enzyme Blends replace traditional collagenase, which is a crude and variable fermentation by-product of Clostridium histolyticum.[3]

Other Notes

  • This product is intended for life science research and in vitro use only. These products are not to be used for diagnostic or clinical applications, such as human islet transplantation.
  • All 1st generation Liberase and Liberase Blendzyme products (Liberase HI, CI, RI, PI, Liberase Blendzyme 1, 2, 3, 4) have been replaced by the 2nd generation Liberase Research Grade and Liberase GMP grade enzyme blend portfolios. For instructions concerning the transition from previously used Liberase Enzymes

For life science research only. Not for use in diagnostic procedures.

Preparation Note

Stabilizers: Calcium
Working concentration: Liberase Research Grade Enzyme Working Concentration

Liberase enzymes have significantly higher specific activities than traditional collagenases. This means that the working concentration of Liberase Research Grade Purified Enzymes, expressed in mg/ml, will be lower than that of traditional collagenase.

When the application is on the Roche list of applications at www.collagenase.com, use the Liberase Research Grade concentration recommended for that application.

When the application is not included on this list, first use Liberase TM Research Grade at a concentration of 0.08–0.28 Wünsch units/ml.

The goal is to determine the best starting concentration of Liberase Research Grade Enzyme Blends. This is a starting point, and the final concentration may vary due to differences in procedure and lot-to-lot differences in traditional collagenase.

Collagenase Working Concentration

Multiply your previous collagenase working concentration (mg/ml) by its specific activity (Wünsch units/mg, [as determined above]), to obtain Wünsch units/ml. To determine how much Liberase Research Grade Enzyme Blend to use, first multiply your collagenase working concentration (in Wünsch units/ml) times the total volume of your working enzyme solution to obtain the total collagenase activity needed (Wünsch units). Divide the total collagenase activity required by the Liberase Research Grade stock concentration ( “Reconstitution and Storage”). This indicates how many milliliters of Liberase Research Grade Enzyme Blend stock solution to use in your working enzyme solution.


Storage conditions (working solution): Store unused stock solution in single-use aliquots at -15 to -25 °C. For further information on product stability, please visit the Roche Liberase Enzyme website at www.collagenase.com.

Note: Avoid repeated freezing and thawing!


Reconstitute the lyophilized enzyme with injection-quality sterile water or tissue-dissociation buffer. Do not add serum or other components (e.g., albumin or protease inhibitors) to the dissociation buffer. Enzyme stability is reduced at higher concentrations and warmer temperatures (4 °C). Avoid both the above conditions.

Reconstitute the entire vial. Do not weigh individual aliquots of the lyophilizate. The introduction of moisture into the vial results in a decline in enzymatic activity.

Place vial on ice to rehydrate the lyophilized enzyme.

Gently agitate the vial at 2 to 8 °C until the enzyme is completely dissolved (max. 30 min).

Depending on the type of tissue-dissociation buffer used to dissolve Liberase Research Grade Purified Enzyme Blends, slight precipitations may be observed which readily dissolve in the diluted working solution, and have no influence on enzyme activity.

Remove an aliquot of the stock solution to prepare the working solution.

Reconstitution volume

2 ml (1 vial with 5 mg–10 mg pack size)
10 ml (1 vial with 50 mg–100 mg pack size)

Collagenase Wünsch (units/ml)

13 (1 vial with 5 mg–10 mg pack size)
26 (1 vial with 50 mg–100 mg pack size)

Total Collagenase concentration [mg/ml]

2.5 (1 vial with 5 mg–10 mg pack size)
5.0 (1 vial with 50 mg–100 mg pack size)
Store dry

Legal Information

Dispase is a registered trademark of Godo Shusei Co., Ltd.
Liberase is a trademark of Roche

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of keratinocytes and fibroblasts from human foreskin by one-step enzyme incubation using liberase research grade products.
Heymer A
Biochemica, 2, 12-14 (2009)
S R L Stacpoole et al.
Cell death and differentiation, 18(6), 1016-1023 (2011-01-29)
In vitro stem cell systems traditionally employ oxygen levels that are far removed from the in vivo situation. This study investigates whether an ambient environment containing a physiological oxygen level of 3% (normoxia) enables the generation of neural precursor cells
Victoria A Elliott et al.
PloS one, 9(5), e97432-e97432 (2014-05-16)
Liver metastasis is the most common cause of death in patients with colorectal cancer. Despite extensive research into the biology of cancer progression, the molecular mechanisms that drive colorectal cancer metastasis are not well characterized. HT29 LM1, HT29 LM2, HT29

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service