11004760001
Roche
Random Primed DNA Labeling Kit
Synonym(s):
nucleic acid labeling
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 50 labeling reactions
Quality Level
manufacturer/tradename
Roche
storage temp.
−20°C
General description
Random primed DNA labeling kit is used for the radioactive and nonradioactive labeling of DNA with modified deoxyribonucleoside triphosphate using random oligonucleotides as primers. In this method, the complementary DNA strand of denatured DNA is synthesized by Klenow polymerase using the 3′-OH termini of the random oligonucleotides as primers.
Specificity
Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0) and/or by heating to 65 °C for 10 minutes.
Application
Random Primed DNA Labeling Kit has been used to label probe or fragments shorter than 1 kb.
Random Primed DNA Labeling Kit is used to uniformly label plasmid or phage DNA with any [α-32P]-dNTP or modified dNTP. Labeled DNA probes with high specific activity are used in a variety of hybridization techniques including:
- Screening of gene libraries
- Southern, dot and northern blots
- In situ hybridizations
Packaging
1 kit containing 7 components.
Preparation Note
Working solution: Preparation dNTP Stock Mix
To avoid pipetting inaccuracy, due to low volumes, prepare a stock mix of unlabeled dNTPs. Aliquots should be stored at -15 to -25 °C. Avoid repeated freezing and thawing.
Preparation of DIG Stock Mix
To avoid pipetting inaccuracy due to low volumes, prepare a DIG stock mix. To do this, mix digoxigenin-11-dUTP [3 mM] and dTTP (vial 5) 1:1. For each labeling reaction 1.6 μl are used.
To avoid pipetting inaccuracy, due to low volumes, prepare a stock mix of unlabeled dNTPs. Aliquots should be stored at -15 to -25 °C. Avoid repeated freezing and thawing.
- dATP, dGTP, dTTP mixture:
For one labeling reaction pipette:
1 μl dATP
1 μl dGTP
1 μl dTTP to a reaction vial
Preparation of DIG Stock Mix
To avoid pipetting inaccuracy due to low volumes, prepare a DIG stock mix. To do this, mix digoxigenin-11-dUTP [3 mM] and dTTP (vial 5) 1:1. For each labeling reaction 1.6 μl are used.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Kit Components Only
Product No.
Description
- Control DNA lambda, 20 μl 12.5 µg/ml
- dATP, 50 μl 0.5 mM
- dCTP, 50 μl 0.5 mM
- dGTP, 50 μl 0.5 mM
- dTTP, 50 μl 0.5 mM
- Hexanucleotide mixture in 10x reaction buffer (100 μl)
- Klenow enzyme, labeling grade (100 U)
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cell, 167(2), 498-511 (2016-10-04)
During eukaryotic DNA interstrand cross-link (ICL) repair, cross-links are resolved ("unhooked") by nucleolytic incisions surrounding the lesion. In vertebrates, ICL repair is triggered when replication forks collide with the lesion, leading to FANCI-FANCD2-dependent unhooking and formation of a double-strand break
The Journal of biological chemistry, 276(26), 24223-24231 (2001-04-11)
Cells infected with the intracellular protozoan parasite Toxoplasma gondii undergo up-regulation of pro-inflammatory cytokines, organelle redistribution, and protection from apoptosis. To examine the molecular basis of these and other changes, gene expression profiles of human foreskin fibroblasts infected with Toxoplasma
The Journal of biological chemistry, 280(41), 34635-34643 (2005-07-27)
Hyperpolarization-activated, cyclic nucleotide-gated cation (HCN) channels underlie the inward pacemaker current, termed I(f)/I(h), in a variety of tissues. Many details are known for the HCN subtypes 1, 2, and 4. We now successfully cloned the cDNA for HCN3 from human
Fluorescence in situ hybridization to the polytene chromosomes of anopheles mosquitoes
PLoS Pathogens (2012)
Cell cycle (Georgetown, Tex.), 12(13), 2084-2099 (2013-06-14)
Immortal cells require a mechanism of telomere length control in order to divide infinitely. One mechanism is telomerase, an enzyme that compensates the loss of telomeric DNA. The second mechanism is the alternative lengthening of telomeres (ALT) pathway. In ALT
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service